Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 44689617-44689872 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC43
Entrez Gene ID
124808
Gene Name
coiled-coil domain containing 43
Gene Aliases
-
UniGene
Hs.579115
Gene Chromosome Location
Chr.17: 44677437-44689797 on Build GRCh38
Transcripts
NM_001099225.1 NM_144609.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7275432 c.106G>A p.D36N Chr.17: 44689648-44689648 on Build GRCh38
6189946 c.46G>A p.D16N Chr.17: 44689708-44689708 on Build GRCh38
6622222 c.124G>A p.A42T Chr.17: 44689630-44689630 on Build GRCh38
4506812 c.72C>T p.S24S Chr.17: 44689682-44689682 on Build GRCh38
5006136 c.76C>A p.L26M Chr.17: 44689678-44689678 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs932993099 Chr.17: 44689683-44689683 on Build GRCh38
rs577156843 Chr.17: 44689777-44689777 on Build GRCh38
rs571125561 Chr.17: 44689692-44689692 on Build GRCh38
rs780839933 Chr.17: 44689722-44689722 on Build GRCh38
rs780470085 Chr.17: 44689702-44689713 on Build GRCh38
rs754435777 Chr.17: 44689694-44689694 on Build GRCh38
rs764629245 Chr.17: 44689854-44689854 on Build GRCh38
rs377376704 Chr.17: 44689669-44689669 on Build GRCh38
rs774964391 Chr.17: 44689672-44689672 on Build GRCh38
rs574488486 Chr.17: 44689810-44689810 on Build GRCh38
rs751879483 Chr.17: 44689783-44689783 on Build GRCh38
rs1034895079 Chr.17: 44689765-44689765 on Build GRCh38
rs762640158 Chr.17: 44689676-44689676 on Build GRCh38
rs760130904 Chr.17: 44689762-44689762 on Build GRCh38
rs751171178 Chr.17: 44689691-44689691 on Build GRCh38
rs752795490 Chr.17: 44689707-44689707 on Build GRCh38
rs774005920 Chr.17: 44689734-44689734 on Build GRCh38
rs758568460 Chr.17: 44689771-44689771 on Build GRCh38
rs778165854 Chr.17: 44689712-44689712 on Build GRCh38
rs969245907 Chr.17: 44689843-44689843 on Build GRCh38
rs1025721992 Chr.17: 44689758-44689758 on Build GRCh38
rs779183794 Chr.17: 44689699-44689699 on Build GRCh38
rs534943447 Chr.17: 44689729-44689729 on Build GRCh38
rs768104210 Chr.17: 44689733-44689733 on Build GRCh38
rs769693933 Chr.17: 44689723-44689723 on Build GRCh38
rs757402961 Chr.17: 44689801-44689801 on Build GRCh38
rs763703326 Chr.17: 44689682-44689682 on Build GRCh38
rs761304877 Chr.17: 44689742-44689742 on Build GRCh38
rs898648128 Chr.17: 44689756-44689756 on Build GRCh38
rs762103999 Chr.17: 44689685-44689685 on Build GRCh38
rs758515134 Chr.17: 44689708-44689708 on Build GRCh38
rs988084983 Chr.17: 44689804-44689804 on Build GRCh38
rs1005688570 Chr.17: 44689725-44689725 on Build GRCh38
rs375264066 Chr.17: 44689644-44689644 on Build GRCh38
rs758648263 Chr.17: 44689694-44689695 on Build GRCh38
rs747145048 Chr.17: 44689716-44689716 on Build GRCh38
rs968173697 Chr.17: 44689845-44689845 on Build GRCh38
rs765878212 Chr.17: 44689764-44689764 on Build GRCh38
rs775019345 Chr.17: 44689726-44689726 on Build GRCh38
rs755428500 Chr.17: 44689698-44689698 on Build GRCh38
rs770767662 Chr.17: 44689720-44689720 on Build GRCh38
rs199713872 Chr.17: 44689642-44689642 on Build GRCh38
rs200562682 Chr.17: 44689651-44689651 on Build GRCh38
rs113179032 Chr.17: 44689757-44689757 on Build GRCh38
rs753208664 Chr.17: 44689766-44689766 on Build GRCh38
rs944985712 Chr.17: 44689645-44689645 on Build GRCh38
rs766825168 Chr.17: 44689745-44689745 on Build GRCh38

View Full Product Details