Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 28336568-28336837 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AZI2
Entrez Gene ID
64343
Gene Name
5-azacytidine induced 2
Gene Aliases
AZ2, NAP1, TILP
UniGene
Hs.706676
Gene Chromosome Location
Chr.3: 28322353-28349129 on Build GRCh38
Transcripts
NM_001134432.1 NM_001134433.1 NM_001271650.1 NM_022461.4 XM_005265392.2 XM_011534009.2 XM_011534010.2 XM_011534011.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4840132 c.553G>A p.D185N Chr.3: 28336772-28336772 on Build GRCh38
4840131 c.553G>A p.D185N Chr.3: 28336772-28336772 on Build GRCh38
6846554 c.529G>C p.E177Q Chr.3: 28336796-28336796 on Build GRCh38
6846555 c.529G>C p.E177Q Chr.3: 28336796-28336796 on Build GRCh38
4638304 c.552C>T p.S184S Chr.3: 28336773-28336773 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs9855649 Chr.3: 28336692-28336692 on Build GRCh38
rs778060510 Chr.3: 28336742-28336742 on Build GRCh38
rs531293273 Chr.3: 28336619-28336619 on Build GRCh38
rs768918380 Chr.3: 28336816-28336816 on Build GRCh38
rs933802343 Chr.3: 28336777-28336777 on Build GRCh38
rs777831982 Chr.3: 28336711-28336711 on Build GRCh38
rs372293222 Chr.3: 28336728-28336728 on Build GRCh38
rs201211825 Chr.3: 28336772-28336772 on Build GRCh38
rs760864525 Chr.3: 28336797-28336797 on Build GRCh38
rs979848763 Chr.3: 28336608-28336608 on Build GRCh38
rs977485623 Chr.3: 28336648-28336648 on Build GRCh38
rs759384904 Chr.3: 28336730-28336730 on Build GRCh38
rs925933234 Chr.3: 28336674-28336674 on Build GRCh38
rs749362794 Chr.3: 28336783-28336784 on Build GRCh38
rs967076448 Chr.3: 28336636-28336636 on Build GRCh38
rs754437353 Chr.3: 28336723-28336723 on Build GRCh38
rs369336325 Chr.3: 28336738-28336738 on Build GRCh38
rs916962720 Chr.3: 28336621-28336621 on Build GRCh38
rs751085171 Chr.3: 28336713-28336713 on Build GRCh38
rs992238124 Chr.3: 28336684-28336684 on Build GRCh38
rs546565020 Chr.3: 28336664-28336664 on Build GRCh38
rs1048763786 Chr.3: 28336802-28336802 on Build GRCh38
rs779511461 Chr.3: 28336770-28336770 on Build GRCh38
rs771573457 Chr.3: 28336755-28336755 on Build GRCh38
rs923902991 Chr.3: 28336612-28336612 on Build GRCh38
rs544375524 Chr.3: 28336602-28336602 on Build GRCh38
rs533309466 Chr.3: 28336628-28336628 on Build GRCh38
rs749596216 Chr.3: 28336749-28336749 on Build GRCh38

View Full Product Details