Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 26312026-26312251 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRF3
Entrez Gene ID
165082
Gene Name
adhesion G protein-coupled receptor F3
Gene Aliases
GPR113, PGR23
UniGene
Hs.631878
Gene Chromosome Location
Chr.2: 26308173-26346817 on Build GRCh38
Transcripts
NM_001145168.1 NM_001145169.1 NM_001321971.1 NM_001321975.1 NM_153835.3 NR_135916.1 XM_011532621.2 XM_011532622.2 XM_011532623.2 XM_017003505.1 XM_017003506.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4583414 c.1101G>A p.R367R Chr.2: 26312030-26312030 on Build GRCh38
6673496 c.1672G>T p.D558Y Chr.2: 26312056-26312056 on Build GRCh38
6673497 c.1075G>T p.D359Y Chr.2: 26312056-26312056 on Build GRCh38
4583413 c.1698G>A p.R566R Chr.2: 26312030-26312030 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs974979629 Chr.2: 26312135-26312135 on Build GRCh38
rs1013563747 Chr.2: 26312221-26312221 on Build GRCh38
rs546671211 Chr.2: 26312131-26312131 on Build GRCh38
rs754706376 Chr.2: 26312069-26312069 on Build GRCh38
rs895167151 Chr.2: 26312197-26312197 on Build GRCh38
rs376993125 Chr.2: 26312122-26312122 on Build GRCh38
rs747692574 Chr.2: 26312100-26312100 on Build GRCh38
rs987696043 Chr.2: 26312188-26312188 on Build GRCh38
rs772321386 Chr.2: 26312103-26312103 on Build GRCh38
rs542072054 Chr.2: 26312072-26312072 on Build GRCh38
rs1055196967 Chr.2: 26312183-26312183 on Build GRCh38
rs762099837 Chr.2: 26312120-26312120 on Build GRCh38
rs745940609 Chr.2: 26312100-26312100 on Build GRCh38
rs779289612 Chr.2: 26312091-26312091 on Build GRCh38
rs769076297 Chr.2: 26312114-26312114 on Build GRCh38
rs751537084 Chr.2: 26312068-26312068 on Build GRCh38
rs903678809 Chr.2: 26312171-26312171 on Build GRCh38
rs776989660 Chr.2: 26312119-26312119 on Build GRCh38
rs115663836 Chr.2: 26312073-26312073 on Build GRCh38
rs528524514 Chr.2: 26312130-26312130 on Build GRCh38
rs112825174 Chr.2: 26312113-26312113 on Build GRCh38
rs78752370 Chr.2: 26312099-26312099 on Build GRCh38
rs375200413 Chr.2: 26312104-26312104 on Build GRCh38
rs1042171165 Chr.2: 26312148-26312148 on Build GRCh38
rs757431614 Chr.2: 26312087-26312087 on Build GRCh38
rs77295721 Chr.2: 26312182-26312182 on Build GRCh38

View Full Product Details