Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 118344331-118344576 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD3G
Entrez Gene ID
917
Gene Name
CD3g molecule
Gene Aliases
CD3-GAMMA, IMD17, T3G
UniGene
Hs.2259
Gene Chromosome Location
Chr.11: 118344307-118353782 on Build GRCh38
Transcripts
NM_000073.2 XM_005271724.3 XM_006718941.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1704617 c.3G>A p.M1I Chr.11: 118344426-118344426 on Build GRCh38
6552713 c.18C>T p.G6G Chr.11: 118344441-118344441 on Build GRCh38
4670496 c.27delC p.L10fs*35 Chr.11: 118344450-118344450 on Build GRCh38
3718429 c.10G>A p.G4R Chr.11: 118344433-118344433 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs375928479 Chr.11: 118344418-118344418 on Build GRCh38
rs145293635 Chr.11: 118344407-118344407 on Build GRCh38
rs1011444458 Chr.11: 118344397-118344397 on Build GRCh38
rs768957947 Chr.11: 118344494-118344494 on Build GRCh38
rs377262966 Chr.11: 118344463-118344463 on Build GRCh38
rs200728539 Chr.11: 118344469-118344469 on Build GRCh38
rs60810919 Chr.11: 118344378-118344381 on Build GRCh38
rs866223443 Chr.11: 118344451-118344451 on Build GRCh38
rs893001013 Chr.11: 118344359-118344359 on Build GRCh38
rs993869702 Chr.11: 118344362-118344362 on Build GRCh38
rs140349750 Chr.11: 118344387-118344387 on Build GRCh38
rs776993949 Chr.11: 118344500-118344500 on Build GRCh38
rs202202803 Chr.11: 118344364-118344364 on Build GRCh38
rs200326844 Chr.11: 118344398-118344398 on Build GRCh38
rs370839678 Chr.11: 118344498-118344498 on Build GRCh38
rs749243996 Chr.11: 118344415-118344415 on Build GRCh38
rs746667725 Chr.11: 118344476-118344476 on Build GRCh38
rs104894199 Chr.11: 118344424-118344424 on Build GRCh38
rs533613241 Chr.11: 118344515-118344515 on Build GRCh38
rs201276257 Chr.11: 118344405-118344405 on Build GRCh38
rs201912617 Chr.11: 118344516-118344516 on Build GRCh38
rs201006956 Chr.11: 118344455-118344455 on Build GRCh38
rs200933474 Chr.11: 118344513-118344513 on Build GRCh38
rs529225294 Chr.11: 118344400-118344400 on Build GRCh38
rs201508267 Chr.11: 118344423-118344423 on Build GRCh38
rs181713283 Chr.11: 118344459-118344459 on Build GRCh38
rs200107110 Chr.11: 118344496-118344496 on Build GRCh38
rs370352015 Chr.11: 118344433-118344433 on Build GRCh38
rs201786349 Chr.11: 118344479-118344479 on Build GRCh38
rs867432113 Chr.11: 118344432-118344432 on Build GRCh38
rs150376137 Chr.11: 118344361-118344364 on Build GRCh38
rs538018516 Chr.11: 118344420-118344420 on Build GRCh38
rs372956565 Chr.11: 118344392-118344392 on Build GRCh38
rs143990986 Chr.11: 118344456-118344456 on Build GRCh38
rs762228493 Chr.11: 118344507-118344507 on Build GRCh38
rs199553749 Chr.11: 118344406-118344406 on Build GRCh38
rs200390526 Chr.11: 118344519-118344519 on Build GRCh38

View Full Product Details