Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 75350690-75350937 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HMGCR
Entrez Gene ID
3156
Gene Name
3-hydroxy-3-methylglutaryl-CoA reductase
Gene Aliases
LDLCQ3
UniGene
Hs.628096
Gene Chromosome Location
Chr.5: 75336334-75362101 on Build GRCh38
Transcripts
NM_000859.2 NM_001130996.1 XM_011543357.1 XM_011543358.1 XM_011543359.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
74931 c.829C>G p.P277A Chr.5: 75350837-75350837 on Build GRCh38
3617871 c.917C>T p.S306F Chr.5: 75350925-75350925 on Build GRCh38
7193409 c.784C>G p.L262V Chr.5: 75350792-75350792 on Build GRCh38
1070101 c.899G>T p.R300I Chr.5: 75350907-75350907 on Build GRCh38
6901203 c.913G>A p.V305I Chr.5: 75350921-75350921 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1029103328 Chr.5: 75350916-75350916 on Build GRCh38
rs377514952 Chr.5: 75350814-75350814 on Build GRCh38
rs139776831 Chr.5: 75350870-75350870 on Build GRCh38
rs781640240 Chr.5: 75350801-75350801 on Build GRCh38
rs565156124 Chr.5: 75350887-75350887 on Build GRCh38
rs754778341 Chr.5: 75350752-75350752 on Build GRCh38
rs776658461 Chr.5: 75350857-75350857 on Build GRCh38
rs1041407285 Chr.5: 75350722-75350722 on Build GRCh38
rs906811306 Chr.5: 75350824-75350824 on Build GRCh38
rs770038652 Chr.5: 75350834-75350834 on Build GRCh38
rs1051258640 Chr.5: 75350831-75350831 on Build GRCh38
rs376236389 Chr.5: 75350753-75350753 on Build GRCh38
rs759558565 Chr.5: 75350865-75350865 on Build GRCh38
rs752336438 Chr.5: 75350761-75350761 on Build GRCh38
rs998709032 Chr.5: 75350809-75350809 on Build GRCh38
rs554100316 Chr.5: 75350841-75350843 on Build GRCh38
rs188568526 Chr.5: 75350745-75350745 on Build GRCh38
rs953163444 Chr.5: 75350917-75350917 on Build GRCh38
rs941371482 Chr.5: 75350728-75350728 on Build GRCh38
rs752430097 Chr.5: 75350885-75350885 on Build GRCh38
rs1009799375 Chr.5: 75350854-75350854 on Build GRCh38
rs770089730 Chr.5: 75350807-75350807 on Build GRCh38
rs199517638 Chr.5: 75350785-75350785 on Build GRCh38
rs763628844 Chr.5: 75350888-75350888 on Build GRCh38
rs973447586 Chr.5: 75350899-75350899 on Build GRCh38
rs544999212 Chr.5: 75350835-75350835 on Build GRCh38
rs746504798 Chr.5: 75350794-75350794 on Build GRCh38
rs756709926 Chr.5: 75350905-75350905 on Build GRCh38
rs542306053 Chr.5: 75350735-75350735 on Build GRCh38
rs113025572 Chr.5: 75350812-75350812 on Build GRCh38
rs751250779 Chr.5: 75350896-75350896 on Build GRCh38
rs144780232 Chr.5: 75350838-75350838 on Build GRCh38
rs1014962118 Chr.5: 75350851-75350851 on Build GRCh38
rs768747516 Chr.5: 75350823-75350823 on Build GRCh38
rs968339885 Chr.5: 75350876-75350876 on Build GRCh38
rs866151085 Chr.5: 75350746-75350746 on Build GRCh38
rs767738315 Chr.5: 75350852-75350852 on Build GRCh38
rs1025150103 Chr.5: 75350822-75350822 on Build GRCh38
rs1055477224 Chr.5: 75350791-75350791 on Build GRCh38
rs757991730 Chr.5: 75350778-75350778 on Build GRCh38
rs774684544 Chr.5: 75350837-75350837 on Build GRCh38

View Full Product Details