Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49890762-49891035 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL4I1
Entrez Gene ID
259307
Gene Name
interleukin 4 induced 1
Gene Aliases
FIG1, LAAO, LAO
UniGene
Hs.574492
Gene Chromosome Location
Chr.19: 49889654-49929539 on Build GRCh38
Transcripts
NM_001258017.1 NM_001258018.1 NM_152899.1 NM_172374.2 NR_047577.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6378835 c.767G>T p.R256I Chr.19: 49890977-49890977 on Build GRCh38
4637385 c.804C>A p.A268A Chr.19: 49891006-49891006 on Build GRCh38
6366415 c.823C>A p.L275I Chr.19: 49890987-49890987 on Build GRCh38
6367217 c.824T>G p.L275R Chr.19: 49890986-49890986 on Build GRCh38
6378834 c.833G>T p.R278I Chr.19: 49890977-49890977 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746235700 Chr.19: 49890948-49890957 on Build GRCh38
rs140238257 Chr.19: 49891012-49891012 on Build GRCh38
rs62126284 Chr.19: 49890956-49890956 on Build GRCh38
rs369590509 Chr.19: 49890891-49890891 on Build GRCh38
rs915437423 Chr.19: 49890918-49890918 on Build GRCh38
rs533936001 Chr.19: 49890939-49890939 on Build GRCh38
rs781612489 Chr.19: 49890934-49890934 on Build GRCh38
rs544885147 Chr.19: 49890946-49890946 on Build GRCh38
rs778151936 Chr.19: 49890957-49890957 on Build GRCh38
rs998542533 Chr.19: 49890857-49890857 on Build GRCh38
rs773198707 Chr.19: 49890937-49890938 on Build GRCh38
rs749963823 Chr.19: 49890948-49890948 on Build GRCh38
rs1054043205 Chr.19: 49890787-49890787 on Build GRCh38
rs568487723 Chr.19: 49890829-49890829 on Build GRCh38
rs765960953 Chr.19: 49890949-49890949 on Build GRCh38
rs774109586 Chr.19: 49890956-49890957 on Build GRCh38
rs752853214 Chr.19: 49890945-49890948 on Build GRCh38
rs973682644 Chr.19: 49890890-49890890 on Build GRCh38
rs966161500 Chr.19: 49890837-49890837 on Build GRCh38
rs66537733 Chr.19: 49890947-49890948 on Build GRCh38
rs962593371 Chr.19: 49890883-49890883 on Build GRCh38
rs753665651 Chr.19: 49890937-49890943 on Build GRCh38
rs749775396 Chr.19: 49890961-49890961 on Build GRCh38
rs1018625647 Chr.19: 49890833-49890833 on Build GRCh38
rs777685535 Chr.19: 49890946-49890948 on Build GRCh38
rs1009500215 Chr.19: 49890863-49890863 on Build GRCh38
rs766921720 Chr.19: 49890937-49890937 on Build GRCh38
rs558862694 Chr.19: 49890942-49890942 on Build GRCh38
rs1031410168 Chr.19: 49890869-49890869 on Build GRCh38
rs572843531 Chr.19: 49890938-49890938 on Build GRCh38
rs773809204 Chr.19: 49890936-49890948 on Build GRCh38
rs939364198 Chr.19: 49890938-49890939 on Build GRCh38
rs770968694 Chr.19: 49890941-49890941 on Build GRCh38
rs754663127 Chr.19: 49890940-49890948 on Build GRCh38
rs1007494456 Chr.19: 49890985-49890985 on Build GRCh38
rs889041772 Chr.19: 49890799-49890799 on Build GRCh38
rs769995693 Chr.19: 49890953-49890953 on Build GRCh38
rs917334159 Chr.19: 49890940-49890940 on Build GRCh38
rs781748723 Chr.19: 49890948-49890949 on Build GRCh38
rs554609847 Chr.19: 49890936-49890936 on Build GRCh38
rs760405621 Chr.19: 49890943-49890943 on Build GRCh38
rs899548378 Chr.19: 49890805-49890805 on Build GRCh38
rs550479324 Chr.19: 49890793-49890793 on Build GRCh38
rs56091826 Chr.19: 49890947-49890947 on Build GRCh38
rs761398462 Chr.19: 49890936-49890936 on Build GRCh38
rs757819273 Chr.19: 49890962-49890962 on Build GRCh38
rs747749462 Chr.19: 49890957-49890958 on Build GRCh38
rs981159822 Chr.19: 49890928-49890928 on Build GRCh38
rs772283932 Chr.19: 49890988-49890988 on Build GRCh38
rs758573815 Chr.19: 49890944-49890945 on Build GRCh38
rs996516504 Chr.19: 49890806-49890806 on Build GRCh38
rs113068493 Chr.19: 49890945-49890945 on Build GRCh38
rs958697687 Chr.19: 49890931-49890931 on Build GRCh38
rs776623486 Chr.19: 49891006-49891006 on Build GRCh38
rs760586038 Chr.19: 49890936-49890937 on Build GRCh38
rs746782149 Chr.19: 49890995-49890995 on Build GRCh38
rs768382889 Chr.19: 49890930-49890930 on Build GRCh38
rs930423704 Chr.19: 49890946-49890947 on Build GRCh38
rs889002728 Chr.19: 49890978-49890979 on Build GRCh38
rs112330118 Chr.19: 49890944-49890944 on Build GRCh38
rs1029377664 Chr.19: 49890814-49890814 on Build GRCh38
rs766257232 Chr.19: 49890937-49890944 on Build GRCh38
rs62126285 Chr.19: 49890958-49890958 on Build GRCh38
rs56080840 Chr.19: 49890925-49890925 on Build GRCh38
rs1027651801 Chr.19: 49890923-49890923 on Build GRCh38
rs745943528 Chr.19: 49890978-49890978 on Build GRCh38
rs563195646 Chr.19: 49890994-49890994 on Build GRCh38
rs753054353 Chr.19: 49890953-49890953 on Build GRCh38
rs748465354 Chr.19: 49890935-49890935 on Build GRCh38
rs757858545 Chr.19: 49890948-49890948 on Build GRCh38
rs1015115767 Chr.19: 49890881-49890881 on Build GRCh38
rs765514611 Chr.19: 49890943-49890944 on Build GRCh38

View Full Product Details