Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 227733241-227733492 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JMJD4
Entrez Gene ID
65094
Gene Name
jumonji domain containing 4
Gene Aliases
-
UniGene
Hs.555974
Gene Chromosome Location
Chr.1: 227731189-227735423 on Build GRCh38
Transcripts
NM_001161465.1 NM_023007.2 XM_011544262.2 NM_001323930.1 NM_001323932.1 NM_001323933.1 NR_136654.1 XM_017000231.1 XM_017000232.1

Gene Symbol
SNAP47
Entrez Gene ID
116841
Gene Name
synaptosome associated protein 47
Gene Aliases
C1orf142, ESFI5812, HEL-S-290, HEL170, SNAP-47, SVAP1
UniGene
Hs.325081
Gene Chromosome Location
Chr.1: 227728168-227781231 on Build GRCh38
Transcripts
NM_001161465.1 NM_023007.2 XM_011544262.2 NM_001323930.1 NM_001323932.1 NM_001323933.1 NR_136654.1 XM_017000231.1 XM_017000232.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3356527 c.903G>C p.Q301H Chr.1: 227733471-227733471 on Build GRCh38
4693855 c.931A>G p.S311G Chr.1: 227733443-227733443 on Build GRCh38
4693856 c.899C>T p.T300M Chr.1: 227733475-227733475 on Build GRCh38
7274199 c.955A>G p.N319D Chr.1: 227733419-227733419 on Build GRCh38
4562075 c.910G>A p.G304S Chr.1: 227733464-227733464 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750894986 Chr.1: 227733370-227733370 on Build GRCh38
rs766897994 Chr.1: 227733474-227733474 on Build GRCh38
rs897146690 Chr.1: 227733389-227733389 on Build GRCh38
rs761688620 Chr.1: 227733274-227733274 on Build GRCh38
rs767941859 Chr.1: 227733363-227733363 on Build GRCh38
rs746902443 Chr.1: 227733409-227733409 on Build GRCh38
rs547932751 Chr.1: 227733450-227733450 on Build GRCh38
rs779171320 Chr.1: 227733392-227733392 on Build GRCh38
rs978616596 Chr.1: 227733362-227733362 on Build GRCh38
rs772845815 Chr.1: 227733406-227733406 on Build GRCh38
rs760950775 Chr.1: 227733378-227733378 on Build GRCh38
rs775085027 Chr.1: 227733443-227733443 on Build GRCh38
rs1008461035 Chr.1: 227733336-227733336 on Build GRCh38
rs758656398 Chr.1: 227733300-227733300 on Build GRCh38
rs766761701 Chr.1: 227733384-227733384 on Build GRCh38
rs547648599 Chr.1: 227733264-227733264 on Build GRCh38
rs41270161 Chr.1: 227733261-227733261 on Build GRCh38
rs1001585426 Chr.1: 227733461-227733461 on Build GRCh38
rs749049017 Chr.1: 227733383-227733384 on Build GRCh38
rs769406665 Chr.1: 227733434-227733434 on Build GRCh38
rs375726420 Chr.1: 227733295-227733295 on Build GRCh38
rs370394993 Chr.1: 227733276-227733276 on Build GRCh38
rs766826979 Chr.1: 227733381-227733381 on Build GRCh38
rs948600024 Chr.1: 227733454-227733454 on Build GRCh38
rs370323713 Chr.1: 227733391-227733391 on Build GRCh38
rs112625101 Chr.1: 227733267-227733267 on Build GRCh38
rs114123047 Chr.1: 227733308-227733308 on Build GRCh38
rs758450961 Chr.1: 227733395-227733395 on Build GRCh38
rs954690069 Chr.1: 227733338-227733338 on Build GRCh38
rs964928641 Chr.1: 227733294-227733294 on Build GRCh38
rs761230709 Chr.1: 227733466-227733466 on Build GRCh38
rs780955014 Chr.1: 227733425-227733425 on Build GRCh38
rs762392178 Chr.1: 227733449-227733449 on Build GRCh38
rs148217485 Chr.1: 227733465-227733465 on Build GRCh38
rs745572833 Chr.1: 227733428-227733428 on Build GRCh38
rs752852960 Chr.1: 227733394-227733394 on Build GRCh38
rs370662794 Chr.1: 227733422-227733422 on Build GRCh38
rs768156108 Chr.1: 227733462-227733462 on Build GRCh38

View Full Product Details