Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 226994101-226994368 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC42BPA
Entrez Gene ID
8476
Gene Name
CDC42 binding protein kinase alpha
Gene Aliases
MRCK, MRCKA, PK428
UniGene
Hs.35433
Gene Chromosome Location
Chr.1: 226989865-227318492 on Build GRCh38
Transcripts
NM_003607.3 NM_014826.4 XM_005273317.3 XM_005273318.3 XM_005273320.3 XM_005273321.3 XM_005273322.3 XM_005273324.3 XM_006711834.3 XM_006711835.3 XM_011544306.2 XM_011544307.2 XM_011544308.2 XM_011544309.2 XM_017002578.1 XM_017002579.1 XM_017002580.1 XM_017002581.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7336196 c.4912C>T p.P1638S Chr.1: 226994273-226994273 on Build GRCh38
7336197 c.5341C>T p.P1781S Chr.1: 226994273-226994273 on Build GRCh38
6624342 c.5233G>T p.E1745* Chr.1: 226994300-226994300 on Build GRCh38
6624341 c.5314G>T p.E1772* Chr.1: 226994300-226994300 on Build GRCh38
6624344 c.4885G>T p.E1629* Chr.1: 226994300-226994300 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs537202979 Chr.1: 226994122-226994122 on Build GRCh38
rs1013205564 Chr.1: 226994152-226994152 on Build GRCh38
rs752901736 Chr.1: 226994220-226994220 on Build GRCh38
rs917515448 Chr.1: 226994308-226994308 on Build GRCh38
rs751642687 Chr.1: 226994303-226994303 on Build GRCh38
rs777019251 Chr.1: 226994341-226994341 on Build GRCh38
rs369925265 Chr.1: 226994224-226994224 on Build GRCh38
rs555455666 Chr.1: 226994285-226994285 on Build GRCh38
rs1015891475 Chr.1: 226994171-226994171 on Build GRCh38
rs988829523 Chr.1: 226994217-226994217 on Build GRCh38
rs2802269 Chr.1: 226994332-226994332 on Build GRCh38
rs772531780 Chr.1: 226994180-226994180 on Build GRCh38
rs896908975 Chr.1: 226994160-226994160 on Build GRCh38
rs373581147 Chr.1: 226994288-226994288 on Build GRCh38
rs768336948 Chr.1: 226994325-226994325 on Build GRCh38
rs142402392 Chr.1: 226994281-226994281 on Build GRCh38
rs896066292 Chr.1: 226994140-226994140 on Build GRCh38
rs890930120 Chr.1: 226994151-226994151 on Build GRCh38
rs373832771 Chr.1: 226994342-226994342 on Build GRCh38
rs56027641 Chr.1: 226994271-226994271 on Build GRCh38
rs866655178 Chr.1: 226994201-226994201 on Build GRCh38
rs545978830 Chr.1: 226994330-226994330 on Build GRCh38
rs765625128 Chr.1: 226994218-226994218 on Build GRCh38
rs764383159 Chr.1: 226994302-226994302 on Build GRCh38
rs965250898 Chr.1: 226994199-226994199 on Build GRCh38
rs768177493 Chr.1: 226994243-226994243 on Build GRCh38
rs144798693 Chr.1: 226994237-226994237 on Build GRCh38
rs752767194 Chr.1: 226994327-226994328 on Build GRCh38
rs369792934 Chr.1: 226994287-226994287 on Build GRCh38
rs979079395 Chr.1: 226994286-226994286 on Build GRCh38
rs869029351 Chr.1: 226994341-226994342 on Build GRCh38
rs985690392 Chr.1: 226994185-226994185 on Build GRCh38
rs995462742 Chr.1: 226994184-226994184 on Build GRCh38
rs3795449 Chr.1: 226994137-226994137 on Build GRCh38
rs371666866 Chr.1: 226994236-226994236 on Build GRCh38
rs200292265 Chr.1: 226994244-226994244 on Build GRCh38
rs748969764 Chr.1: 226994324-226994324 on Build GRCh38
rs146538430 Chr.1: 226994311-226994311 on Build GRCh38
rs748877740 Chr.1: 226994239-226994239 on Build GRCh38
rs3795452 Chr.1: 226994245-226994245 on Build GRCh38
rs995294242 Chr.1: 226994176-226994176 on Build GRCh38
rs1023028273 Chr.1: 226994189-226994189 on Build GRCh38
rs570776494 Chr.1: 226994155-226994155 on Build GRCh38
rs776041416 Chr.1: 226994346-226994346 on Build GRCh38
rs946389435 Chr.1: 226994273-226994273 on Build GRCh38
rs3795450 Chr.1: 226994159-226994159 on Build GRCh38
rs376204917 Chr.1: 226994310-226994310 on Build GRCh38
rs1019550018 Chr.1: 226994188-226994188 on Build GRCh38
rs778513565 Chr.1: 226994327-226994327 on Build GRCh38
rs770318162 Chr.1: 226994345-226994345 on Build GRCh38
rs145018031 Chr.1: 226994323-226994323 on Build GRCh38
rs757185096 Chr.1: 226994234-226994234 on Build GRCh38
rs757348849 Chr.1: 226994309-226994309 on Build GRCh38
rs3795451 Chr.1: 226994222-226994222 on Build GRCh38
rs781297756 Chr.1: 226994232-226994236 on Build GRCh38
rs772011182 Chr.1: 226994142-226994142 on Build GRCh38

View Full Product Details