Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 44194714-44194939 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATXN7L3
Entrez Gene ID
56970
Gene Name
ataxin 7 like 3
Gene Aliases
-
UniGene
Hs.512651
Gene Chromosome Location
Chr.17: 44191805-44199976 on Build GRCh38
Transcripts
NM_001098833.1 NM_020218.1 XM_011525038.2 XM_011525044.2 XM_011525046.2 XM_017024880.1 XM_017024881.1 XM_017024882.1 XM_017024883.1 XM_017024884.1 XM_017024885.1 XM_017024886.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1383621 c.703C>T p.R235* Chr.17: 44194802-44194802 on Build GRCh38
7273748 c.730G>A p.V244I Chr.17: 44194796-44194796 on Build GRCh38
7129960 c.726C>A p.L242L Chr.17: 44194779-44194779 on Build GRCh38
4784005 c.695G>A p.R232H Chr.17: 44194831-44194831 on Build GRCh38
7129959 c.747C>A p.L249L Chr.17: 44194779-44194779 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146231360 Chr.17: 44194797-44194797 on Build GRCh38
rs6503491 Chr.17: 44194870-44194870 on Build GRCh38
rs757647507 Chr.17: 44194785-44194785 on Build GRCh38
rs564777185 Chr.17: 44194779-44194779 on Build GRCh38
rs182223869 Chr.17: 44194890-44194890 on Build GRCh38
rs375442756 Chr.17: 44194754-44194754 on Build GRCh38
rs773302205 Chr.17: 44194761-44194761 on Build GRCh38
rs772004617 Chr.17: 44194848-44194848 on Build GRCh38
rs779310372 Chr.17: 44194832-44194832 on Build GRCh38
rs753356109 Chr.17: 44194769-44194769 on Build GRCh38
rs376690555 Chr.17: 44194882-44194882 on Build GRCh38
rs749385874 Chr.17: 44194736-44194736 on Build GRCh38
rs750678946 Chr.17: 44194860-44194860 on Build GRCh38
rs181092765 Chr.17: 44194770-44194770 on Build GRCh38
rs774693921 Chr.17: 44194748-44194748 on Build GRCh38
rs368897355 Chr.17: 44194759-44194759 on Build GRCh38
rs768819005 Chr.17: 44194740-44194740 on Build GRCh38
rs552529939 Chr.17: 44194803-44194803 on Build GRCh38
rs756479733 Chr.17: 44194887-44194887 on Build GRCh38
rs756649643 Chr.17: 44194801-44194801 on Build GRCh38
rs375374643 Chr.17: 44194778-44194778 on Build GRCh38
rs771955420 Chr.17: 44194757-44194757 on Build GRCh38
rs779936033 Chr.17: 44194734-44194734 on Build GRCh38
rs766458110 Chr.17: 44194881-44194881 on Build GRCh38
rs34990731 Chr.17: 44194781-44194782 on Build GRCh38
rs76721333 Chr.17: 44194873-44194873 on Build GRCh38
rs372309757 Chr.17: 44194773-44194773 on Build GRCh38
rs201555978 Chr.17: 44194849-44194849 on Build GRCh38
rs764591254 Chr.17: 44194739-44194739 on Build GRCh38
rs765918316 Chr.17: 44194768-44194768 on Build GRCh38
rs767331959 Chr.17: 44194837-44194837 on Build GRCh38
rs186812360 Chr.17: 44194836-44194836 on Build GRCh38
rs749377744 Chr.17: 44194824-44194824 on Build GRCh38
rs759005197 Chr.17: 44194868-44194868 on Build GRCh38
rs901443247 Chr.17: 44194892-44194892 on Build GRCh38
rs756363472 Chr.17: 44194865-44194865 on Build GRCh38
rs371315935 Chr.17: 44194857-44194857 on Build GRCh38
rs781480220 Chr.17: 44194796-44194796 on Build GRCh38
rs532831353 Chr.17: 44194907-44194907 on Build GRCh38
rs760513365 Chr.17: 44194851-44194851 on Build GRCh38

View Full Product Details