Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49659447-49659718 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IRF3
Entrez Gene ID
3661
Gene Name
interferon regulatory factor 3
Gene Aliases
IIAE7
UniGene
Hs.289052
Gene Chromosome Location
Chr.19: 49659569-49665875 on Build GRCh38
Transcripts
NM_001197122.1 NM_001197123.1 NM_001197124.1 NM_001197125.1 NM_001197126.1 NM_001197127.1 NM_001197128.1 NM_001571.5 NR_045568.1 XM_006723197.1 XM_006723198.1 XM_006723200.1 XM_006723201.1 XM_006723202.2 XM_017026766.1 XM_017026767.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6366020 c.1262T>A p.F421Y Chr.19: 49659670-49659670 on Build GRCh38
1727608 c.1258delG p.D420fs*>8 Chr.19: 49659674-49659674 on Build GRCh38
2753019 c.1276G>A p.E426K Chr.19: 49659656-49659656 on Build GRCh38
1471240 c.1215C>T p.S405S Chr.19: 49659717-49659717 on Build GRCh38
148687 c.1280G>C p.S427T Chr.19: 49659652-49659652 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs767690928 Chr.19: 49659582-49659582 on Build GRCh38
rs778636886 Chr.19: 49659590-49659590 on Build GRCh38
rs769042253 Chr.19: 49659636-49659636 on Build GRCh38
rs748641459 Chr.19: 49659656-49659656 on Build GRCh38
rs55642332 Chr.19: 49659553-49659554 on Build GRCh38
rs759848637 Chr.19: 49659578-49659578 on Build GRCh38
rs368936131 Chr.19: 49659663-49659663 on Build GRCh38
rs768284312 Chr.19: 49659622-49659622 on Build GRCh38
rs199920869 Chr.19: 49659641-49659641 on Build GRCh38
rs549087839 Chr.19: 49659503-49659503 on Build GRCh38
rs941749156 Chr.19: 49659631-49659631 on Build GRCh38
rs201693357 Chr.19: 49659616-49659616 on Build GRCh38
rs756652776 Chr.19: 49659613-49659613 on Build GRCh38
rs776238308 Chr.19: 49659629-49659629 on Build GRCh38
rs750476278 Chr.19: 49659613-49659614 on Build GRCh38
rs758135685 Chr.19: 49659615-49659615 on Build GRCh38
rs746513741 Chr.19: 49659545-49659545 on Build GRCh38
rs746196443 Chr.19: 49659613-49659613 on Build GRCh38
rs924825996 Chr.19: 49659608-49659608 on Build GRCh38
rs891660841 Chr.19: 49659490-49659490 on Build GRCh38
rs373306574 Chr.19: 49659619-49659619 on Build GRCh38
rs767689722 Chr.19: 49659611-49659612 on Build GRCh38
rs7251 Chr.19: 49659652-49659652 on Build GRCh38
rs746779352 Chr.19: 49659472-49659472 on Build GRCh38
rs754292336 Chr.19: 49659642-49659642 on Build GRCh38
rs757832899 Chr.19: 49659689-49659689 on Build GRCh38
rs201177412 Chr.19: 49659642-49659642 on Build GRCh38
rs752845931 Chr.19: 49659600-49659600 on Build GRCh38
rs576101389 Chr.19: 49659640-49659640 on Build GRCh38
rs141490768 Chr.19: 49659696-49659696 on Build GRCh38
rs1010053237 Chr.19: 49659499-49659499 on Build GRCh38
rs753813852 Chr.19: 49659688-49659688 on Build GRCh38
rs748135462 Chr.19: 49659634-49659634 on Build GRCh38
rs765109901 Chr.19: 49659569-49659569 on Build GRCh38
rs567985173 Chr.19: 49659532-49659532 on Build GRCh38
rs764192287 Chr.19: 49659664-49659664 on Build GRCh38
rs757666757 Chr.19: 49659609-49659609 on Build GRCh38
rs370098271 Chr.19: 49659618-49659618 on Build GRCh38
rs372047994 Chr.19: 49659601-49659601 on Build GRCh38
rs966352964 Chr.19: 49659524-49659524 on Build GRCh38
rs1007519280 Chr.19: 49659495-49659495 on Build GRCh38
rs772830437 Chr.19: 49659572-49659572 on Build GRCh38
rs903906778 Chr.19: 49659507-49659507 on Build GRCh38
rs756176484 Chr.19: 49659603-49659603 on Build GRCh38
rs779357935 Chr.19: 49659612-49659612 on Build GRCh38
rs1001282847 Chr.19: 49659515-49659515 on Build GRCh38
rs138524983 Chr.19: 49659555-49659555 on Build GRCh38
rs796264933 Chr.19: 49659613-49659614 on Build GRCh38
rs978062551 Chr.19: 49659542-49659542 on Build GRCh38
rs775706485 Chr.19: 49659658-49659658 on Build GRCh38
rs891691722 Chr.19: 49659674-49659674 on Build GRCh38
rs776721308 Chr.19: 49659561-49659561 on Build GRCh38
rs754349455 Chr.19: 49659604-49659604 on Build GRCh38

View Full Product Details