Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 74295723-74295948 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC33
Entrez Gene ID
80125
Gene Name
coiled-coil domain containing 33
Gene Aliases
CT61, HP11097
UniGene
Hs.383206
Gene Chromosome Location
Chr.15: 74202705-74336472 on Build GRCh38
Transcripts
NM_025055.4 XM_005254692.1 XM_011522085.2 XM_011522086.1 XM_011522087.2 XM_011522088.1 XM_011522089.2 XM_017022623.1 XM_017022624.1 XM_017022625.1 XM_017022626.1 XM_017022627.1 XM_017022628.1 XM_017022629.1 XM_017022630.1 XM_017022631.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4151130 c.1212C>T p.S404S Chr.15: 74295870-74295870 on Build GRCh38
6439170 c.1199A>G p.K400R Chr.15: 74295857-74295857 on Build GRCh38
434276 c.1247A>G p.E416G Chr.15: 74295905-74295905 on Build GRCh38
6852937 c.1801T>C p.W601R Chr.15: 74295850-74295850 on Build GRCh38
5590087 c.1767C>T p.I589I Chr.15: 74295816-74295816 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774758633 Chr.15: 74295892-74295896 on Build GRCh38
rs779739810 Chr.15: 74295869-74295869 on Build GRCh38
rs547023567 Chr.15: 74295814-74295814 on Build GRCh38
rs1040834328 Chr.15: 74295913-74295913 on Build GRCh38
rs199650392 Chr.15: 74295847-74295847 on Build GRCh38
rs924160630 Chr.15: 74295885-74295885 on Build GRCh38
rs776863953 Chr.15: 74295927-74295927 on Build GRCh38
rs117429899 Chr.15: 74295891-74295891 on Build GRCh38
rs772176628 Chr.15: 74295918-74295918 on Build GRCh38
rs142120320 Chr.15: 74295791-74295791 on Build GRCh38
rs773487437 Chr.15: 74295768-74295768 on Build GRCh38
rs761890622 Chr.15: 74295749-74295749 on Build GRCh38
rs774401065 Chr.15: 74295900-74295900 on Build GRCh38
rs922010566 Chr.15: 74295829-74295829 on Build GRCh38
rs773251731 Chr.15: 74295919-74295919 on Build GRCh38
rs190768916 Chr.15: 74295856-74295856 on Build GRCh38
rs749075303 Chr.15: 74295879-74295879 on Build GRCh38
rs748070126 Chr.15: 74295902-74295902 on Build GRCh38
rs565232246 Chr.15: 74295750-74295750 on Build GRCh38
rs760907164 Chr.15: 74295920-74295920 on Build GRCh38
rs375321958 Chr.15: 74295793-74295793 on Build GRCh38
rs369347846 Chr.15: 74295813-74295813 on Build GRCh38
rs755470888 Chr.15: 74295800-74295800 on Build GRCh38
rs1054054658 Chr.15: 74295890-74295890 on Build GRCh38
rs766546171 Chr.15: 74295925-74295925 on Build GRCh38
rs978859153 Chr.15: 74295887-74295887 on Build GRCh38
rs756886208 Chr.15: 74295840-74295840 on Build GRCh38
rs551793242 Chr.15: 74295888-74295888 on Build GRCh38
rs960249029 Chr.15: 74295839-74295839 on Build GRCh38
rs1035963431 Chr.15: 74295801-74295801 on Build GRCh38
rs374164076 Chr.15: 74295821-74295821 on Build GRCh38
rs745558245 Chr.15: 74295854-74295854 on Build GRCh38

View Full Product Details