Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27368317-27368564 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF2B4
Entrez Gene ID
8890
Gene Name
eukaryotic translation initiation factor 2B subunit delta
Gene Aliases
EIF-2B, EIF2B, EIF2Bdelta
UniGene
Hs.169474
Gene Chromosome Location
Chr.2: 27364352-27370457 on Build GRCh38
Transcripts
NM_001034116.1 NM_001318965.1 NM_001318966.1 NM_001318967.1 NM_001318968.1 NM_001318969.1 NM_015636.3 NM_172195.3 XM_006712132.1 XM_011533147.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1641896 c.526A>G p.K176E Chr.2: 27368436-27368436 on Build GRCh38
3839280 c.544C>T p.H182Y Chr.2: 27368418-27368418 on Build GRCh38
6333237 c.558C>A p.Y186* Chr.2: 27368404-27368404 on Build GRCh38
6273524 c.584C>A p.S195Y Chr.2: 27368438-27368438 on Build GRCh38
6316290 c.565C>A p.Q189K Chr.2: 27368397-27368397 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs944661678 Chr.2: 27368511-27368511 on Build GRCh38
rs566287769 Chr.2: 27368495-27368495 on Build GRCh38
rs769763044 Chr.2: 27368474-27368474 on Build GRCh38
rs746547024 Chr.2: 27368457-27368457 on Build GRCh38
rs879271655 Chr.2: 27368415-27368415 on Build GRCh38
rs913265804 Chr.2: 27368538-27368538 on Build GRCh38
rs776656241 Chr.2: 27368350-27368350 on Build GRCh38
rs547695043 Chr.2: 27368489-27368489 on Build GRCh38
rs375458330 Chr.2: 27368396-27368396 on Build GRCh38
rs752818834 Chr.2: 27368379-27368379 on Build GRCh38
rs757657538 Chr.2: 27368416-27368416 on Build GRCh38
rs565245777 Chr.2: 27368427-27368427 on Build GRCh38
rs776743727 Chr.2: 27368467-27368467 on Build GRCh38
rs756090304 Chr.2: 27368392-27368392 on Build GRCh38
rs758641876 Chr.2: 27368535-27368535 on Build GRCh38
rs374326886 Chr.2: 27368516-27368516 on Build GRCh38
rs911814858 Chr.2: 27368498-27368498 on Build GRCh38
rs370256543 Chr.2: 27368513-27368513 on Build GRCh38
rs761726266 Chr.2: 27368351-27368351 on Build GRCh38
rs886055902 Chr.2: 27368447-27368447 on Build GRCh38
rs779502336 Chr.2: 27368444-27368444 on Build GRCh38
rs758061891 Chr.2: 27368443-27368443 on Build GRCh38
rs990980109 Chr.2: 27368361-27368361 on Build GRCh38
rs754390217 Chr.2: 27368509-27368509 on Build GRCh38
rs370123021 Chr.2: 27368479-27368479 on Build GRCh38
rs768638665 Chr.2: 27368346-27368346 on Build GRCh38
rs373696834 Chr.2: 27368352-27368352 on Build GRCh38
rs377475649 Chr.2: 27368493-27368493 on Build GRCh38
rs764126847 Chr.2: 27368506-27368506 on Build GRCh38
rs772991139 Chr.2: 27368390-27368390 on Build GRCh38
rs745575196 Chr.2: 27368430-27368430 on Build GRCh38
rs748069510 Chr.2: 27368469-27368469 on Build GRCh38
rs928893380 Chr.2: 27368500-27368500 on Build GRCh38
rs373303281 Chr.2: 27368488-27368488 on Build GRCh38
rs191734334 Chr.2: 27368355-27368355 on Build GRCh38
rs920428422 Chr.2: 27368439-27368439 on Build GRCh38
rs767553031 Chr.2: 27368375-27368375 on Build GRCh38
rs1053334796 Chr.2: 27368540-27368540 on Build GRCh38

View Full Product Details