Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 63027068-63027341 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GGH
Entrez Gene ID
8836
Gene Name
gamma-glutamyl hydrolase
Gene Aliases
GH
UniGene
Hs.78619
Gene Chromosome Location
Chr.8: 63015079-63039051 on Build GRCh38
Transcripts
NM_003878.2 XM_011517623.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6670694 c.326A>T p.K109I Chr.8: 63027215-63027215 on Build GRCh38
346935 c.323C>G p.A108G Chr.8: 63027218-63027218 on Build GRCh38
1314142 c.306C>G p.L102L Chr.8: 63027235-63027235 on Build GRCh38
336957 c.289G>T p.G97* Chr.8: 63027252-63027252 on Build GRCh38
6036186 c.333C>T p.A111A Chr.8: 63027208-63027208 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770885990 Chr.8: 63027235-63027235 on Build GRCh38
rs543685028 Chr.8: 63027311-63027311 on Build GRCh38
rs762971905 Chr.8: 63027135-63027135 on Build GRCh38
rs191194248 Chr.8: 63027304-63027304 on Build GRCh38
rs528348849 Chr.8: 63027231-63027231 on Build GRCh38
rs982288243 Chr.8: 63027122-63027122 on Build GRCh38
rs377550396 Chr.8: 63027309-63027309 on Build GRCh38
rs148095972 Chr.8: 63027313-63027313 on Build GRCh38
rs747742633 Chr.8: 63027303-63027303 on Build GRCh38
rs961541147 Chr.8: 63027106-63027106 on Build GRCh38
rs147030498 Chr.8: 63027126-63027126 on Build GRCh38
rs147855753 Chr.8: 63027250-63027250 on Build GRCh38
rs545671872 Chr.8: 63027202-63027202 on Build GRCh38
rs915252649 Chr.8: 63027225-63027225 on Build GRCh38
rs377603060 Chr.8: 63027275-63027275 on Build GRCh38
rs758953408 Chr.8: 63027182-63027182 on Build GRCh38
rs938299609 Chr.8: 63027127-63027127 on Build GRCh38
rs901476590 Chr.8: 63027171-63027171 on Build GRCh38
rs756435325 Chr.8: 63027312-63027313 on Build GRCh38
rs1040026220 Chr.8: 63027140-63027140 on Build GRCh38
rs780793720 Chr.8: 63027215-63027215 on Build GRCh38
rs750906276 Chr.8: 63027174-63027174 on Build GRCh38
rs757367589 Chr.8: 63027163-63027163 on Build GRCh38
rs564440258 Chr.8: 63027230-63027230 on Build GRCh38
rs547411787 Chr.8: 63027237-63027237 on Build GRCh38
rs774302773 Chr.8: 63027312-63027312 on Build GRCh38
rs2305558 Chr.8: 63027280-63027280 on Build GRCh38
rs370611780 Chr.8: 63027276-63027276 on Build GRCh38
rs541905158 Chr.8: 63027097-63027097 on Build GRCh38
rs759342215 Chr.8: 63027239-63027239 on Build GRCh38
rs372364012 Chr.8: 63027296-63027296 on Build GRCh38
rs769245235 Chr.8: 63027220-63027220 on Build GRCh38
rs781559752 Chr.8: 63027299-63027299 on Build GRCh38
rs936113311 Chr.8: 63027107-63027107 on Build GRCh38
rs747830409 Chr.8: 63027218-63027218 on Build GRCh38
rs759306005 Chr.8: 63027160-63027160 on Build GRCh38
rs758468500 Chr.8: 63027287-63027287 on Build GRCh38
rs780496107 Chr.8: 63027186-63027186 on Build GRCh38
rs766020779 Chr.8: 63027146-63027146 on Build GRCh38
rs776940313 Chr.8: 63027253-63027253 on Build GRCh38
rs200731720 Chr.8: 63027166-63027166 on Build GRCh38
rs777460035 Chr.8: 63027307-63027307 on Build GRCh38
rs1054668599 Chr.8: 63027115-63027115 on Build GRCh38
rs766890450 Chr.8: 63027294-63027294 on Build GRCh38
rs747389650 Chr.8: 63027187-63027187 on Build GRCh38
rs754150665 Chr.8: 63027162-63027162 on Build GRCh38
rs751365620 Chr.8: 63027157-63027157 on Build GRCh38

View Full Product Details