Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 24785573-24785846 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GMNN
Entrez Gene ID
51053
Gene Name
geminin, DNA replication inhibitor
Gene Aliases
Gem, MGORS6
UniGene
Hs.234896
Gene Chromosome Location
Chr.6: 24774931-24786099 on Build GRCh38
Transcripts
NM_001251989.1 NM_001251990.1 NM_001251991.1 NM_015895.4 XM_005249159.1 XM_011514651.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6573831 c.481G>C p.E161Q Chr.6: 24785650-24785650 on Build GRCh38
5221810 c.567_568delAG p.E190fs*6 Chr.6: 24785736-24785737 on Build GRCh38
1076525 c.479G>A p.G160D Chr.6: 24785648-24785648 on Build GRCh38
2856862 c.579G>A p.T193T Chr.6: 24785748-24785748 on Build GRCh38
6479958 c.597A>G p.V199V Chr.6: 24785766-24785766 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775945443 Chr.6: 24785732-24785732 on Build GRCh38
rs777983756 Chr.6: 24785676-24785676 on Build GRCh38
rs780708194 Chr.6: 24785809-24785809 on Build GRCh38
rs906420047 Chr.6: 24785744-24785744 on Build GRCh38
rs761528197 Chr.6: 24785608-24785608 on Build GRCh38
rs781107737 Chr.6: 24785800-24785800 on Build GRCh38
rs773171916 Chr.6: 24785698-24785700 on Build GRCh38
rs756646787 Chr.6: 24785808-24785808 on Build GRCh38
rs745980330 Chr.6: 24785803-24785803 on Build GRCh38
rs758203415 Chr.6: 24785790-24785790 on Build GRCh38
rs991501358 Chr.6: 24785661-24785661 on Build GRCh38
rs141209716 Chr.6: 24785670-24785670 on Build GRCh38
rs749698337 Chr.6: 24785741-24785741 on Build GRCh38
rs375293626 Chr.6: 24785624-24785624 on Build GRCh38
rs781391678 Chr.6: 24785710-24785710 on Build GRCh38
rs760045613 Chr.6: 24785763-24785763 on Build GRCh38
rs753702835 Chr.6: 24785634-24785634 on Build GRCh38
rs369845176 Chr.6: 24785713-24785713 on Build GRCh38
rs2307303 Chr.6: 24785777-24785777 on Build GRCh38
rs773007360 Chr.6: 24785747-24785747 on Build GRCh38
rs558038900 Chr.6: 24785791-24785791 on Build GRCh38
rs767312956 Chr.6: 24785611-24785611 on Build GRCh38
rs765975961 Chr.6: 24785632-24785632 on Build GRCh38
rs771779829 Chr.6: 24785746-24785746 on Build GRCh38
rs747044114 Chr.6: 24785677-24785677 on Build GRCh38
rs201229321 Chr.6: 24785635-24785635 on Build GRCh38
rs775521672 Chr.6: 24785778-24785778 on Build GRCh38
rs770471324 Chr.6: 24785727-24785727 on Build GRCh38
rs778646277 Chr.6: 24785665-24785665 on Build GRCh38
rs373551106 Chr.6: 24785748-24785748 on Build GRCh38
rs758565804 Chr.6: 24785672-24785672 on Build GRCh38
rs776256972 Chr.6: 24785756-24785756 on Build GRCh38
rs776047687 Chr.6: 24785607-24785607 on Build GRCh38
rs766435020 Chr.6: 24785750-24785751 on Build GRCh38
rs949333405 Chr.6: 24785684-24785684 on Build GRCh38
rs562408220 Chr.6: 24785705-24785705 on Build GRCh38
rs757280891 Chr.6: 24785694-24785694 on Build GRCh38
rs939630843 Chr.6: 24785709-24785710 on Build GRCh38
rs757538616 Chr.6: 24785797-24785797 on Build GRCh38
rs796867391 Chr.6: 24785771-24785771 on Build GRCh38
rs759345110 Chr.6: 24785773-24785773 on Build GRCh38
rs749906997 Chr.6: 24785623-24785623 on Build GRCh38
rs753847779 Chr.6: 24785814-24785817 on Build GRCh38
rs771954624 Chr.6: 24785642-24785645 on Build GRCh38

View Full Product Details