Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 45153775-45154026 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXorf36
Entrez Gene ID
79742
Gene Name
chromosome X open reading frame 36
Gene Aliases
4930578C19Rik, DIA1R, EPQL1862, PRO3743, bA435K1.1
UniGene
Hs.98321
Gene Chromosome Location
Chr.X: 45148373-45200901 on Build GRCh38
Transcripts
NM_176819.3 XM_005272670.1 XM_006724559.1

SNP Detail

SNP ID Chromosome Location
rs770173706 Chr.X: 45153900-45153900 on Build GRCh38
rs376081979 Chr.X: 45153941-45153941 on Build GRCh38
rs113373165 Chr.X: 45153816-45153816 on Build GRCh38
rs778769796 Chr.X: 45153976-45153976 on Build GRCh38
rs771521366 Chr.X: 45153979-45153979 on Build GRCh38
rs963701029 Chr.X: 45153984-45153984 on Build GRCh38
rs757066033 Chr.X: 45153965-45153965 on Build GRCh38
rs758115140 Chr.X: 45153923-45153925 on Build GRCh38
rs775053239 Chr.X: 45153884-45153884 on Build GRCh38
rs759584364 Chr.X: 45153946-45153946 on Build GRCh38
rs1037071483 Chr.X: 45153986-45153986 on Build GRCh38
rs1032686828 Chr.X: 45153982-45153982 on Build GRCh38
rs139672027 Chr.X: 45153803-45153803 on Build GRCh38
rs770877997 Chr.X: 45153858-45153858 on Build GRCh38
rs777721060 Chr.X: 45153958-45153958 on Build GRCh38
rs766864510 Chr.X: 45153928-45153928 on Build GRCh38
rs6611072 Chr.X: 45153852-45153852 on Build GRCh38
rs184665277 Chr.X: 45153827-45153827 on Build GRCh38
rs763456706 Chr.X: 45153923-45153923 on Build GRCh38
rs745794862 Chr.X: 45153977-45153977 on Build GRCh38
rs767607729 Chr.X: 45153947-45153947 on Build GRCh38
rs189703445 Chr.X: 45153921-45153921 on Build GRCh38
rs779575269 Chr.X: 45153980-45153980 on Build GRCh38
rs975022120 Chr.X: 45153996-45153996 on Build GRCh38
rs753003946 Chr.X: 45153948-45153948 on Build GRCh38
rs896762018 Chr.X: 45153856-45153856 on Build GRCh38
rs756358717 Chr.X: 45153949-45153949 on Build GRCh38
rs760289781 Chr.X: 45153895-45153895 on Build GRCh38
rs771778814 Chr.X: 45153810-45153810 on Build GRCh38
rs1057130525 Chr.X: 45153840-45153840 on Build GRCh38
rs746684634 Chr.X: 45153987-45153987 on Build GRCh38
rs371509825 Chr.X: 45153964-45153964 on Build GRCh38
rs1045450355 Chr.X: 45153859-45153859 on Build GRCh38

View Full Product Details