Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 118312022-118312295 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD3E
Entrez Gene ID
916
Gene Name
CD3e molecule
Gene Aliases
IMD18, T3E, TCRE
UniGene
Hs.3003
Gene Chromosome Location
Chr.11: 118304580-118316175 on Build GRCh38
Transcripts
NM_000733.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6623753 c.86-1G>T p.? Chr.11: 118312152-118312152 on Build GRCh38
5985828 c.103+5G>A p.? Chr.11: 118312175-118312175 on Build GRCh38
6415749 c.101C>A p.T34K Chr.11: 118312168-118312168 on Build GRCh38
3687016 c.93T>C p.I31I Chr.11: 118312160-118312160 on Build GRCh38
6131231 c.103C>T p.P35S Chr.11: 118312170-118312170 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs200719182 Chr.11: 118312216-118312216 on Build GRCh38
rs1945765 Chr.11: 118312114-118312114 on Build GRCh38
rs778170251 Chr.11: 118312199-118312199 on Build GRCh38
rs778981649 Chr.11: 118312113-118312113 on Build GRCh38
rs202089395 Chr.11: 118312123-118312123 on Build GRCh38
rs199922230 Chr.11: 118312110-118312110 on Build GRCh38
rs200911795 Chr.11: 118312146-118312146 on Build GRCh38
rs910799392 Chr.11: 118312225-118312225 on Build GRCh38
rs980972848 Chr.11: 118312272-118312272 on Build GRCh38
rs1945764 Chr.11: 118312086-118312086 on Build GRCh38
rs191862547 Chr.11: 118312095-118312095 on Build GRCh38
rs201300645 Chr.11: 118312219-118312219 on Build GRCh38
rs969460426 Chr.11: 118312244-118312245 on Build GRCh38
rs773135734 Chr.11: 118312155-118312155 on Build GRCh38
rs767097407 Chr.11: 118312191-118312191 on Build GRCh38
rs377014131 Chr.11: 118312182-118312182 on Build GRCh38
rs202079083 Chr.11: 118312156-118312156 on Build GRCh38
rs752441095 Chr.11: 118312197-118312197 on Build GRCh38
rs754145102 Chr.11: 118312200-118312200 on Build GRCh38
rs201538012 Chr.11: 118312205-118312205 on Build GRCh38
rs200358909 Chr.11: 118312202-118312202 on Build GRCh38
rs775733747 Chr.11: 118312140-118312140 on Build GRCh38
rs146167864 Chr.11: 118312268-118312268 on Build GRCh38
rs914949858 Chr.11: 118312247-118312247 on Build GRCh38
rs369702351 Chr.11: 118312206-118312206 on Build GRCh38
rs143949187 Chr.11: 118312170-118312170 on Build GRCh38
rs200448338 Chr.11: 118312139-118312139 on Build GRCh38
rs762016302 Chr.11: 118312242-118312242 on Build GRCh38
rs199766139 Chr.11: 118312212-118312212 on Build GRCh38
rs570072109 Chr.11: 118312096-118312096 on Build GRCh38
rs758824766 Chr.11: 118312271-118312271 on Build GRCh38
rs201543770 Chr.11: 118312171-118312171 on Build GRCh38
rs200265584 Chr.11: 118312198-118312198 on Build GRCh38
rs987700725 Chr.11: 118312224-118312224 on Build GRCh38
rs765526064 Chr.11: 118312246-118312246 on Build GRCh38
rs538984599 Chr.11: 118312107-118312107 on Build GRCh38
rs760787198 Chr.11: 118312145-118312145 on Build GRCh38
rs143015515 Chr.11: 118312180-118312180 on Build GRCh38

View Full Product Details