Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 108896755-108897006 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPSM2
Entrez Gene ID
29899
Gene Name
G-protein signaling modulator 2
Gene Aliases
CMCS, DFNB82, LGN, PINS
UniGene
Hs.584901
Gene Chromosome Location
Chr.1: 108875350-108934335 on Build GRCh38
Transcripts
NM_001321038.1 NM_001321039.1 NM_013296.4 XM_006710589.2 XM_011541301.2 XM_011541302.2 XM_011541303.2 XM_017001097.1 XM_017001098.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
893410 c.97C>T p.R33C Chr.1: 108896904-108896904 on Build GRCh38
5439458 c.187G>A p.A63T Chr.1: 108896994-108896994 on Build GRCh38
6120140 c.138C>G p.F46L Chr.1: 108896945-108896945 on Build GRCh38
6120141 c.130G>A p.V44M Chr.1: 108896937-108896937 on Build GRCh38
3700444 c.154C>A p.Q52K Chr.1: 108896961-108896961 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762581956 Chr.1: 108896928-108896928 on Build GRCh38
rs900826622 Chr.1: 108896793-108896793 on Build GRCh38
rs781015812 Chr.1: 108896820-108896820 on Build GRCh38
rs755804651 Chr.1: 108896945-108896945 on Build GRCh38
rs916602684 Chr.1: 108896936-108896936 on Build GRCh38
rs201572171 Chr.1: 108896826-108896826 on Build GRCh38
rs767840543 Chr.1: 108896901-108896901 on Build GRCh38
rs764931658 Chr.1: 108896904-108896904 on Build GRCh38
rs771251133 Chr.1: 108896875-108896875 on Build GRCh38
rs777024358 Chr.1: 108896886-108896886 on Build GRCh38
rs752223693 Chr.1: 108896909-108896909 on Build GRCh38
rs749508795 Chr.1: 108896825-108896825 on Build GRCh38
rs748331991 Chr.1: 108896961-108896961 on Build GRCh38
rs761125854 Chr.1: 108896856-108896856 on Build GRCh38
rs545129382 Chr.1: 108896788-108896788 on Build GRCh38
rs1012841600 Chr.1: 108896937-108896937 on Build GRCh38
rs112935966 Chr.1: 108896894-108896894 on Build GRCh38
rs753253217 Chr.1: 108896830-108896831 on Build GRCh38
rs779202698 Chr.1: 108896947-108896947 on Build GRCh38
rs751596044 Chr.1: 108896929-108896929 on Build GRCh38
rs772166294 Chr.1: 108896962-108896962 on Build GRCh38
rs75774985 Chr.1: 108896778-108896778 on Build GRCh38
rs748243964 Chr.1: 108896833-108896833 on Build GRCh38
rs375978544 Chr.1: 108896866-108896866 on Build GRCh38
rs200819801 Chr.1: 108896839-108896839 on Build GRCh38
rs1006621707 Chr.1: 108896781-108896781 on Build GRCh38
rs146120260 Chr.1: 108896779-108896779 on Build GRCh38
rs1038043089 Chr.1: 108896858-108896858 on Build GRCh38
rs777811656 Chr.1: 108896973-108896973 on Build GRCh38
rs756622626 Chr.1: 108896832-108896832 on Build GRCh38
rs745323715 Chr.1: 108896931-108896931 on Build GRCh38
rs982085014 Chr.1: 108896978-108896978 on Build GRCh38
rs778582423 Chr.1: 108896908-108896909 on Build GRCh38
rs978675671 Chr.1: 108896834-108896834 on Build GRCh38
rs753005800 Chr.1: 108896930-108896930 on Build GRCh38
rs370596230 Chr.1: 108896935-108896935 on Build GRCh38
rs778914875 Chr.1: 108896832-108896832 on Build GRCh38
rs780911321 Chr.1: 108896931-108896931 on Build GRCh38
rs572688788 Chr.1: 108896815-108896815 on Build GRCh38
rs184863735 Chr.1: 108896854-108896854 on Build GRCh38
rs966939519 Chr.1: 108896830-108896830 on Build GRCh38
rs997791665 Chr.1: 108896795-108896795 on Build GRCh38

View Full Product Details