Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 150520833-150521104 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GIMAP7
Entrez Gene ID
168537
Gene Name
GTPase, IMAP family member 7
Gene Aliases
IAN7, hIAN7
UniGene
Hs.647074
Gene Chromosome Location
Chr.7: 150514857-150521073 on Build GRCh38
Transcripts
NM_153236.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
746023 c.864G>C p.W288C Chr.7: 150520838-150520838 on Build GRCh38
3698260 c.888G>A p.K296K Chr.7: 150520862-150520862 on Build GRCh38
1622546 c.876G>A p.L292L Chr.7: 150520850-150520850 on Build GRCh38
4587238 c.879G>A p.S293S Chr.7: 150520853-150520853 on Build GRCh38
5531697 c.899C>T p.S300F Chr.7: 150520873-150520873 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs149895065 Chr.7: 150521076-150521076 on Build GRCh38
rs373233332 Chr.7: 150520883-150520883 on Build GRCh38
rs776828644 Chr.7: 150520867-150520868 on Build GRCh38
rs150236188 Chr.7: 150520882-150520882 on Build GRCh38
rs767188719 Chr.7: 150520901-150520901 on Build GRCh38
rs1014043745 Chr.7: 150521004-150521004 on Build GRCh38
rs776827852 Chr.7: 150521075-150521076 on Build GRCh38
rs754925440 Chr.7: 150520919-150520919 on Build GRCh38
rs747143006 Chr.7: 150520869-150520869 on Build GRCh38
rs1012916557 Chr.7: 150521048-150521048 on Build GRCh38
rs895617322 Chr.7: 150520989-150520989 on Build GRCh38
rs776302841 Chr.7: 150521015-150521016 on Build GRCh38
rs946708268 Chr.7: 150520907-150520907 on Build GRCh38
rs750095789 Chr.7: 150520905-150520905 on Build GRCh38
rs77097159 Chr.7: 150520923-150520926 on Build GRCh38
rs902631627 Chr.7: 150520993-150520993 on Build GRCh38
rs141933762 Chr.7: 150520920-150520923 on Build GRCh38
rs776783738 Chr.7: 150520873-150520873 on Build GRCh38
rs776109007 Chr.7: 150520877-150520877 on Build GRCh38
rs61244541 Chr.7: 150521011-150521011 on Build GRCh38
rs765909367 Chr.7: 150520911-150520911 on Build GRCh38
rs886888373 Chr.7: 150521031-150521031 on Build GRCh38
rs1051808821 Chr.7: 150520985-150520985 on Build GRCh38
rs999564346 Chr.7: 150521028-150521029 on Build GRCh38
rs1055544119 Chr.7: 150520938-150520938 on Build GRCh38
rs369111955 Chr.7: 150521029-150521030 on Build GRCh38
rs937299018 Chr.7: 150520937-150520937 on Build GRCh38
rs761302724 Chr.7: 150520895-150520895 on Build GRCh38
rs201633257 Chr.7: 150520923-150520923 on Build GRCh38
rs60687169 Chr.7: 150521030-150521031 on Build GRCh38
rs770041454 Chr.7: 150520876-150520876 on Build GRCh38
rs10578 Chr.7: 150520914-150520914 on Build GRCh38
rs71715597 Chr.7: 150521014-150521015 on Build GRCh38
rs745946130 Chr.7: 150520875-150520875 on Build GRCh38
rs774788971 Chr.7: 150520885-150520885 on Build GRCh38
rs755593437 Chr.7: 150520910-150520910 on Build GRCh38
rs771190654 Chr.7: 150520870-150520870 on Build GRCh38
rs759846991 Chr.7: 150520894-150520894 on Build GRCh38
rs58391005 Chr.7: 150521049-150521049 on Build GRCh38
rs138903749 Chr.7: 150521038-150521038 on Build GRCh38
rs73726888 Chr.7: 150521029-150521029 on Build GRCh38
rs142032527 Chr.7: 150521013-150521014 on Build GRCh38

View Full Product Details