Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 55358617-55358890 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM71E2
Entrez Gene ID
284418
Gene Name
family with sequence similarity 71 member E2
Gene Aliases
C19orf16
UniGene
Hs.528319
Gene Chromosome Location
Chr.19: 55354908-55363252 on Build GRCh38
Transcripts
NM_001145402.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6581335 c.1187C>G p.S396C Chr.19: 55358649-55358649 on Build GRCh38
6495990 c.2172G>A p.L724L Chr.19: 55358696-55358696 on Build GRCh38
7367467 c.2245G>C p.E749Q Chr.19: 55358623-55358623 on Build GRCh38
3396359 c.1080G>A p.T360T Chr.19: 55358756-55358756 on Build GRCh38
3952984 c.1998G>A p.L666L Chr.19: 55358870-55358870 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs923121202 Chr.19: 55358869-55358869 on Build GRCh38
rs982340081 Chr.19: 55358720-55358720 on Build GRCh38
rs368477163 Chr.19: 55358694-55358694 on Build GRCh38
rs185501624 Chr.19: 55358782-55358782 on Build GRCh38
rs768877133 Chr.19: 55358830-55358830 on Build GRCh38
rs12974819 Chr.19: 55358736-55358736 on Build GRCh38
rs564738371 Chr.19: 55358672-55358672 on Build GRCh38
rs764053097 Chr.19: 55358681-55358681 on Build GRCh38
rs193182653 Chr.19: 55358725-55358725 on Build GRCh38
rs866069777 Chr.19: 55358755-55358755 on Build GRCh38
rs539283543 Chr.19: 55358862-55358862 on Build GRCh38
rs990232685 Chr.19: 55358832-55358832 on Build GRCh38
rs780891047 Chr.19: 55358688-55358688 on Build GRCh38
rs543977961 Chr.19: 55358682-55358682 on Build GRCh38
rs549946780 Chr.19: 55358863-55358863 on Build GRCh38
rs138813187 Chr.19: 55358646-55358646 on Build GRCh38
rs923070385 Chr.19: 55358719-55358719 on Build GRCh38
rs1047692851 Chr.19: 55358828-55358828 on Build GRCh38
rs541752025 Chr.19: 55358668-55358668 on Build GRCh38
rs776892496 Chr.19: 55358857-55358857 on Build GRCh38
rs1049821102 Chr.19: 55358735-55358735 on Build GRCh38
rs1001263555 Chr.19: 55358824-55358824 on Build GRCh38
rs374834592 Chr.19: 55358673-55358673 on Build GRCh38
rs747790673 Chr.19: 55358645-55358645 on Build GRCh38
rs912872713 Chr.19: 55358834-55358834 on Build GRCh38
rs945952960 Chr.19: 55358707-55358707 on Build GRCh38
rs766532053 Chr.19: 55358842-55358842 on Build GRCh38
rs750230190 Chr.19: 55358691-55358691 on Build GRCh38
rs767477115 Chr.19: 55358831-55358831 on Build GRCh38
rs148917807 Chr.19: 55358759-55358759 on Build GRCh38
rs751040224 Chr.19: 55358796-55358796 on Build GRCh38
rs578225574 Chr.19: 55358678-55358678 on Build GRCh38
rs769289837 Chr.19: 55358642-55358642 on Build GRCh38
rs769496041 Chr.19: 55358647-55358647 on Build GRCh38
rs773504679 Chr.19: 55358737-55358737 on Build GRCh38
rs369843520 Chr.19: 55358726-55358726 on Build GRCh38
rs756349671 Chr.19: 55358693-55358693 on Build GRCh38
rs1040627442 Chr.19: 55358713-55358713 on Build GRCh38
rs561355442 Chr.19: 55358757-55358757 on Build GRCh38
rs1024442672 Chr.19: 55358783-55358783 on Build GRCh38
rs762387466 Chr.19: 55358679-55358679 on Build GRCh38
rs769340128 Chr.19: 55358756-55358756 on Build GRCh38
rs750198354 Chr.19: 55358835-55358835 on Build GRCh38
rs73607046 Chr.19: 55358701-55358701 on Build GRCh38
rs757167186 Chr.19: 55358687-55358687 on Build GRCh38
rs762288592 Chr.19: 55358661-55358661 on Build GRCh38
rs754160895 Chr.19: 55358844-55358844 on Build GRCh38
rs200752567 Chr.19: 55358820-55358820 on Build GRCh38

View Full Product Details