Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 122937618-122937883 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C10orf88
Entrez Gene ID
80007
Gene Name
chromosome 10 open reading frame 88
Gene Aliases
-
UniGene
Hs.501286
Gene Chromosome Location
Chr.10: 122930903-122954403 on Build GRCh38
Transcripts
NM_024942.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2020850 c.933A>G p.A311A Chr.10: 122937875-122937875 on Build GRCh38
465385 c.955A>T p.K319* Chr.10: 122937853-122937853 on Build GRCh38
5546829 c.1016_1022delGTAGTCA p.C339fs*1 Chr.10: 122937786-122937792 on Build GRCh38
915726 c.994C>A p.L332M Chr.10: 122937814-122937814 on Build GRCh38
6613928 c.992T>C p.L331S Chr.10: 122937816-122937816 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs978623547 Chr.10: 122937695-122937695 on Build GRCh38
rs188159022 Chr.10: 122937784-122937784 on Build GRCh38
rs146772739 Chr.10: 122937818-122937818 on Build GRCh38
rs765185358 Chr.10: 122937823-122937823 on Build GRCh38
rs61751031 Chr.10: 122937756-122937756 on Build GRCh38
rs774944097 Chr.10: 122937786-122937786 on Build GRCh38
rs61751030 Chr.10: 122937754-122937754 on Build GRCh38
rs748832426 Chr.10: 122937780-122937780 on Build GRCh38
rs779792141 Chr.10: 122937775-122937775 on Build GRCh38
rs751415358 Chr.10: 122937751-122937751 on Build GRCh38
rs769217092 Chr.10: 122937688-122937688 on Build GRCh38
rs774652543 Chr.10: 122937714-122937714 on Build GRCh38
rs948480279 Chr.10: 122937771-122937771 on Build GRCh38
rs777773654 Chr.10: 122937748-122937748 on Build GRCh38
rs200811772 Chr.10: 122937686-122937686 on Build GRCh38
rs746837110 Chr.10: 122937665-122937665 on Build GRCh38
rs140611786 Chr.10: 122937820-122937820 on Build GRCh38
rs758349249 Chr.10: 122937766-122937766 on Build GRCh38
rs767724784 Chr.10: 122937721-122937721 on Build GRCh38
rs753977206 Chr.10: 122937746-122937746 on Build GRCh38
rs151291521 Chr.10: 122937728-122937728 on Build GRCh38
rs755903805 Chr.10: 122937857-122937857 on Build GRCh38
rs752802938 Chr.10: 122937829-122937829 on Build GRCh38
rs369169090 Chr.10: 122937674-122937674 on Build GRCh38
rs145403983 Chr.10: 122937755-122937755 on Build GRCh38
rs10794580 Chr.10: 122937682-122937682 on Build GRCh38
rs773634530 Chr.10: 122937807-122937807 on Build GRCh38
rs781258643 Chr.10: 122937839-122937839 on Build GRCh38
rs374851210 Chr.10: 122937667-122937667 on Build GRCh38
rs760927553 Chr.10: 122937739-122937739 on Build GRCh38
rs373306715 Chr.10: 122937675-122937675 on Build GRCh38
rs143584957 Chr.10: 122937795-122937795 on Build GRCh38
rs1033608602 Chr.10: 122937734-122937734 on Build GRCh38
rs777560588 Chr.10: 122937661-122937661 on Build GRCh38
rs140044276 Chr.10: 122937763-122937763 on Build GRCh38
rs772600482 Chr.10: 122937803-122937803 on Build GRCh38
rs750254159 Chr.10: 122937844-122937844 on Build GRCh38
rs776381308 Chr.10: 122937673-122937673 on Build GRCh38
rs976038443 Chr.10: 122937821-122937821 on Build GRCh38
rs759674198 Chr.10: 122937833-122937834 on Build GRCh38
rs761015593 Chr.10: 122937812-122937812 on Build GRCh38
rs758430144 Chr.10: 122937660-122937660 on Build GRCh38
rs200757228 Chr.10: 122937720-122937720 on Build GRCh38
rs762275639 Chr.10: 122937790-122937790 on Build GRCh38
rs766584716 Chr.10: 122937742-122937742 on Build GRCh38
rs574778815 Chr.10: 122937814-122937814 on Build GRCh38
rs375925407 Chr.10: 122937747-122937747 on Build GRCh38

View Full Product Details