Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 152650363-152650632 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRQ
Entrez Gene ID
55879
Gene Name
gamma-aminobutyric acid type A receptor theta subunit
Gene Aliases
THETA
UniGene
Hs.283081
Gene Chromosome Location
Chr.X: 152637895-152659827 on Build GRCh38
Transcripts
NM_018558.3 XM_011531184.2

SNP Detail

SNP ID Chromosome Location
rs782084939 Chr.X: 152650578-152650578 on Build GRCh38
rs781936288 Chr.X: 152650515-152650515 on Build GRCh38
rs376267415 Chr.X: 152650439-152650439 on Build GRCh38
rs782159275 Chr.X: 152650440-152650440 on Build GRCh38
rs782701718 Chr.X: 152650432-152650432 on Build GRCh38
rs782168851 Chr.X: 152650516-152650516 on Build GRCh38
rs35542633 Chr.X: 152650466-152650467 on Build GRCh38
rs782492663 Chr.X: 152650450-152650450 on Build GRCh38
rs782332461 Chr.X: 152650513-152650513 on Build GRCh38
rs782256421 Chr.X: 152650385-152650385 on Build GRCh38
rs782709969 Chr.X: 152650598-152650598 on Build GRCh38
rs781966316 Chr.X: 152650531-152650531 on Build GRCh38
rs781971341 Chr.X: 152650576-152650576 on Build GRCh38
rs782465882 Chr.X: 152650463-152650463 on Build GRCh38
rs782758780 Chr.X: 152650460-152650460 on Build GRCh38
rs782249015 Chr.X: 152650501-152650501 on Build GRCh38
rs782302230 Chr.X: 152650475-152650475 on Build GRCh38
rs781870098 Chr.X: 152650449-152650449 on Build GRCh38
rs782134457 Chr.X: 152650536-152650536 on Build GRCh38
rs781824421 Chr.X: 152650462-152650462 on Build GRCh38
rs782656746 Chr.X: 152650489-152650489 on Build GRCh38
rs782377267 Chr.X: 152650408-152650408 on Build GRCh38
rs782075030 Chr.X: 152650401-152650401 on Build GRCh38
rs373222100 Chr.X: 152650398-152650398 on Build GRCh38
rs782019560 Chr.X: 152650534-152650534 on Build GRCh38
rs782793239 Chr.X: 152650447-152650447 on Build GRCh38
rs782343829 Chr.X: 152650443-152650443 on Build GRCh38
rs782570875 Chr.X: 152650468-152650468 on Build GRCh38
rs782758625 Chr.X: 152650557-152650557 on Build GRCh38
rs782544765 Chr.X: 152650484-152650484 on Build GRCh38

View Full Product Details