Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 67182807-67183072 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD180
Entrez Gene ID
4064
Gene Name
CD180 molecule
Gene Aliases
LY64, Ly78, RP105
UniGene
Hs.87205
Gene Chromosome Location
Chr.5: 67182275-67196789 on Build GRCh38
Transcripts
NM_005582.2 XM_005248504.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3828326 c.1856C>G p.S619C Chr.5: 67182987-67182987 on Build GRCh38
171064 c.1915C>T p.L639L Chr.5: 67182928-67182928 on Build GRCh38
6535615 c.1931T>C p.I644T Chr.5: 67182912-67182912 on Build GRCh38
1634293 c.1808A>G p.E603G Chr.5: 67183035-67183035 on Build GRCh38
4585965 c.1968G>A p.W656* Chr.5: 67182875-67182875 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747699790 Chr.5: 67182916-67182916 on Build GRCh38
rs746362762 Chr.5: 67182985-67182985 on Build GRCh38
rs746684140 Chr.5: 67182975-67182975 on Build GRCh38
rs768796013 Chr.5: 67183001-67183001 on Build GRCh38
rs544924019 Chr.5: 67182983-67182983 on Build GRCh38
rs775679457 Chr.5: 67182988-67182988 on Build GRCh38
rs949324975 Chr.5: 67182885-67182885 on Build GRCh38
rs764261269 Chr.5: 67182844-67182844 on Build GRCh38
rs147445049 Chr.5: 67183037-67183037 on Build GRCh38
rs753770262 Chr.5: 67182899-67182899 on Build GRCh38
rs751830500 Chr.5: 67182866-67182866 on Build GRCh38
rs1024358677 Chr.5: 67182880-67182880 on Build GRCh38
rs777337445 Chr.5: 67183026-67183026 on Build GRCh38
rs771130683 Chr.5: 67182976-67182976 on Build GRCh38
rs1037525068 Chr.5: 67182941-67182941 on Build GRCh38
rs1014350532 Chr.5: 67182876-67182876 on Build GRCh38
rs1017499350 Chr.5: 67182917-67182917 on Build GRCh38
rs770471496 Chr.5: 67182986-67182986 on Build GRCh38
rs918192141 Chr.5: 67182859-67182859 on Build GRCh38
rs766078074 Chr.5: 67183013-67183013 on Build GRCh38
rs1007147644 Chr.5: 67183023-67183023 on Build GRCh38
rs762162192 Chr.5: 67183011-67183011 on Build GRCh38
rs746274461 Chr.5: 67183038-67183038 on Build GRCh38
rs766201593 Chr.5: 67182875-67182875 on Build GRCh38
rs369776137 Chr.5: 67182920-67182920 on Build GRCh38
rs769105461 Chr.5: 67183010-67183010 on Build GRCh38
rs369283882 Chr.5: 67183041-67183041 on Build GRCh38
rs201091750 Chr.5: 67183031-67183031 on Build GRCh38
rs891798277 Chr.5: 67182848-67182848 on Build GRCh38
rs765121474 Chr.5: 67183019-67183019 on Build GRCh38
rs376832414 Chr.5: 67182922-67182922 on Build GRCh38
rs776431180 Chr.5: 67183014-67183014 on Build GRCh38
rs2230524 Chr.5: 67182901-67182901 on Build GRCh38
rs866909606 Chr.5: 67182895-67182895 on Build GRCh38
rs565546385 Chr.5: 67182870-67182870 on Build GRCh38
rs759441508 Chr.5: 67183015-67183015 on Build GRCh38
rs751163709 Chr.5: 67183030-67183030 on Build GRCh38
rs749807841 Chr.5: 67183042-67183042 on Build GRCh38
rs777240115 Chr.5: 67182932-67182932 on Build GRCh38
rs763243563 Chr.5: 67182841-67182841 on Build GRCh38
rs762894875 Chr.5: 67182989-67182989 on Build GRCh38
rs747424517 Chr.5: 67182970-67182970 on Build GRCh38
rs897684612 Chr.5: 67182951-67182951 on Build GRCh38
rs539383079 Chr.5: 67183012-67183013 on Build GRCh38
rs1052094146 Chr.5: 67183008-67183008 on Build GRCh38
rs768039902 Chr.5: 67182881-67182881 on Build GRCh38
rs756092596 Chr.5: 67182954-67182954 on Build GRCh38
rs557132138 Chr.5: 67183025-67183025 on Build GRCh38
rs778376595 Chr.5: 67182904-67182904 on Build GRCh38
rs75456095 Chr.5: 67182961-67182961 on Build GRCh38

View Full Product Details