Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 80620738-80620965 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDYL2
Entrez Gene ID
124359
Gene Name
chromodomain protein, Y-like 2
Gene Aliases
PCCP1
UniGene
Hs.373908
Gene Chromosome Location
Chr.16: 80597899-80805043 on Build GRCh38
Transcripts
NM_152342.3 XM_011522866.1 XM_011522867.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4892791 c.979G>A p.E327K Chr.16: 80620791-80620791 on Build GRCh38
3421184 c.868G>A p.A290T Chr.16: 80620902-80620902 on Build GRCh38
4063040 c.903C>T p.S301S Chr.16: 80620867-80620867 on Build GRCh38
7198439 c.926G>C p.S309T Chr.16: 80620844-80620844 on Build GRCh38
4671300 c.927C>T p.S309S Chr.16: 80620843-80620843 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751494194 Chr.16: 80620814-80620814 on Build GRCh38
rs990171887 Chr.16: 80620879-80620879 on Build GRCh38
rs758460696 Chr.16: 80620909-80620909 on Build GRCh38
rs147605884 Chr.16: 80620921-80620921 on Build GRCh38
rs142002683 Chr.16: 80620787-80620787 on Build GRCh38
rs978792477 Chr.16: 80620924-80620924 on Build GRCh38
rs375436035 Chr.16: 80620942-80620942 on Build GRCh38
rs760496597 Chr.16: 80620896-80620896 on Build GRCh38
rs762507376 Chr.16: 80620808-80620808 on Build GRCh38
rs866902124 Chr.16: 80620866-80620866 on Build GRCh38
rs370310225 Chr.16: 80620782-80620782 on Build GRCh38
rs761655091 Chr.16: 80620815-80620815 on Build GRCh38
rs775181291 Chr.16: 80620875-80620875 on Build GRCh38
rs138973238 Chr.16: 80620786-80620786 on Build GRCh38
rs370746070 Chr.16: 80620890-80620890 on Build GRCh38
rs778486733 Chr.16: 80620854-80620854 on Build GRCh38
rs757864703 Chr.16: 80620920-80620920 on Build GRCh38
rs752955955 Chr.16: 80620906-80620906 on Build GRCh38
rs752769233 Chr.16: 80620840-80620840 on Build GRCh38
rs571363904 Chr.16: 80620804-80620804 on Build GRCh38
rs750237699 Chr.16: 80620895-80620895 on Build GRCh38
rs141862110 Chr.16: 80620867-80620867 on Build GRCh38
rs769385267 Chr.16: 80620871-80620871 on Build GRCh38
rs377532688 Chr.16: 80620885-80620885 on Build GRCh38
rs748983597 Chr.16: 80620933-80620933 on Build GRCh38
rs767400567 Chr.16: 80620891-80620891 on Build GRCh38
rs200676958 Chr.16: 80620912-80620912 on Build GRCh38
rs746263715 Chr.16: 80620923-80620923 on Build GRCh38
rs111324619 Chr.16: 80620910-80620910 on Build GRCh38
rs372002661 Chr.16: 80620941-80620941 on Build GRCh38
rs55968480 Chr.16: 80620778-80620778 on Build GRCh38
rs750153584 Chr.16: 80620822-80620822 on Build GRCh38
rs901339293 Chr.16: 80620807-80620807 on Build GRCh38
rs150794698 Chr.16: 80620881-80620881 on Build GRCh38
rs756500412 Chr.16: 80620928-80620928 on Build GRCh38
rs148507807 Chr.16: 80620843-80620843 on Build GRCh38
rs145308954 Chr.16: 80620903-80620903 on Build GRCh38
rs779182525 Chr.16: 80620803-80620803 on Build GRCh38
rs886091903 Chr.16: 80620870-80620870 on Build GRCh38
rs755541394 Chr.16: 80620759-80620759 on Build GRCh38
rs368202024 Chr.16: 80620913-80620913 on Build GRCh38
rs779145359 Chr.16: 80620832-80620832 on Build GRCh38
rs942854968 Chr.16: 80620758-80620758 on Build GRCh38
rs781716179 Chr.16: 80620864-80620864 on Build GRCh38
rs975671527 Chr.16: 80620934-80620934 on Build GRCh38
rs144737101 Chr.16: 80620809-80620809 on Build GRCh38
rs759272484 Chr.16: 80620797-80620797 on Build GRCh38
rs768141056 Chr.16: 80620882-80620882 on Build GRCh38
rs980430837 Chr.16: 80620893-80620893 on Build GRCh38
rs139799393 Chr.16: 80620897-80620897 on Build GRCh38
rs147914173 Chr.16: 80620820-80620820 on Build GRCh38
rs755185419 Chr.16: 80620824-80620824 on Build GRCh38
rs547188605 Chr.16: 80620855-80620855 on Build GRCh38
rs772478772 Chr.16: 80620768-80620768 on Build GRCh38

View Full Product Details