Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 38326287-38326514 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR179
Entrez Gene ID
440435
Gene Name
G protein-coupled receptor 179
Gene Aliases
CSNB1E, GPR158L, GPR158L1
UniGene
Hs.462915
Gene Chromosome Location
Chr.17: 38325610-38343810 on Build GRCh38
Transcripts
NM_001004334.3

SNP Detail

SNP ID Chromosome Location
rs747538205 Chr.17: 38326416-38326416 on Build GRCh38
rs1026935661 Chr.17: 38326340-38326340 on Build GRCh38
rs951261684 Chr.17: 38326372-38326372 on Build GRCh38
rs763094322 Chr.17: 38326460-38326460 on Build GRCh38
rs373545362 Chr.17: 38326355-38326355 on Build GRCh38
rs768555103 Chr.17: 38326377-38326378 on Build GRCh38
rs995710392 Chr.17: 38326336-38326336 on Build GRCh38
rs980886690 Chr.17: 38326484-38326484 on Build GRCh38
rs982802249 Chr.17: 38326413-38326413 on Build GRCh38
rs868114854 Chr.17: 38326353-38326353 on Build GRCh38
rs971106830 Chr.17: 38326434-38326434 on Build GRCh38
rs541077762 Chr.17: 38326318-38326318 on Build GRCh38
rs990504293 Chr.17: 38326332-38326332 on Build GRCh38
rs746186611 Chr.17: 38326437-38326437 on Build GRCh38
rs775713380 Chr.17: 38326457-38326457 on Build GRCh38
rs944624145 Chr.17: 38326483-38326483 on Build GRCh38
rs58740367 Chr.17: 38326489-38326489 on Build GRCh38
rs771632742 Chr.17: 38326423-38326423 on Build GRCh38
rs927583250 Chr.17: 38326312-38326312 on Build GRCh38
rs79056584 Chr.17: 38326426-38326426 on Build GRCh38
rs770311999 Chr.17: 38326448-38326448 on Build GRCh38
rs768763409 Chr.17: 38326470-38326470 on Build GRCh38
rs914957374 Chr.17: 38326376-38326376 on Build GRCh38

View Full Product Details