Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 24984801-24985058 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENKUR
Entrez Gene ID
219670
Gene Name
enkurin, TRPC channel interacting protein
Gene Aliases
C10orf63, CFAP106
UniGene
Hs.534486
Gene Chromosome Location
Chr.10: 24981979-25062279 on Build GRCh38
Transcripts
NM_001270383.1 NM_145010.3 NR_072992.1 NR_072993.1 XM_006717389.1 XM_006717390.1 XM_011519348.1 XM_017015830.1 XM_017016665.1

Gene Symbol
THNSL1
Entrez Gene ID
79896
Gene Name
threonine synthase like 1
Gene Aliases
TSH1
UniGene
Hs.645274
Gene Chromosome Location
Chr.10: 24952319-25026664 on Build GRCh38
Transcripts
NM_001270383.1 NM_145010.3 NR_072992.1 NR_072993.1 XM_006717389.1 XM_006717390.1 XM_011519348.1 XM_017015830.1 XM_017016665.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2133674 c.628G>A p.E210K Chr.10: 24984872-24984872 on Build GRCh38
4553613 c.596G>A p.G199E Chr.10: 24984904-24984904 on Build GRCh38
4013426 c.690G>A p.L230L Chr.10: 24984810-24984810 on Build GRCh38
6050417 c.621G>A p.V207V Chr.10: 24984879-24984879 on Build GRCh38
7366251 c.685A>G p.R229G Chr.10: 24984815-24984815 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757629109 Chr.10: 24984849-24984849 on Build GRCh38
rs773613333 Chr.10: 24984948-24984948 on Build GRCh38
rs559762509 Chr.10: 24985021-24985021 on Build GRCh38
rs1031366065 Chr.10: 24984856-24984856 on Build GRCh38
rs758998957 Chr.10: 24984836-24984836 on Build GRCh38
rs137882461 Chr.10: 24984828-24984828 on Build GRCh38
rs1011336892 Chr.10: 24984898-24984898 on Build GRCh38
rs770294991 Chr.10: 24984923-24984923 on Build GRCh38
rs77695985 Chr.10: 24984963-24984963 on Build GRCh38
rs765093450 Chr.10: 24984861-24984861 on Build GRCh38
rs749850422 Chr.10: 24984952-24984952 on Build GRCh38
rs529994890 Chr.10: 24984944-24984944 on Build GRCh38
rs957834374 Chr.10: 24985012-24985012 on Build GRCh38
rs747201354 Chr.10: 24984894-24984894 on Build GRCh38
rs564601772 Chr.10: 24984834-24984834 on Build GRCh38
rs906215466 Chr.10: 24984922-24984922 on Build GRCh38
rs750478897 Chr.10: 24984877-24984877 on Build GRCh38
rs200773323 Chr.10: 24984824-24984824 on Build GRCh38
rs763974043 Chr.10: 24984852-24984852 on Build GRCh38
rs752370637 Chr.10: 24984843-24984843 on Build GRCh38
rs115177998 Chr.10: 24984823-24984823 on Build GRCh38
rs757163828 Chr.10: 24984859-24984859 on Build GRCh38
rs755260825 Chr.10: 24984895-24984895 on Build GRCh38
rs758434034 Chr.10: 24984879-24984879 on Build GRCh38
rs764736926 Chr.10: 24984903-24984903 on Build GRCh38
rs141651847 Chr.10: 24984849-24984849 on Build GRCh38
rs201909752 Chr.10: 24984855-24984855 on Build GRCh38
rs561038469 Chr.10: 24984887-24984887 on Build GRCh38
rs575120495 Chr.10: 24984841-24984841 on Build GRCh38

View Full Product Details