Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 13682007-13682232 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM210A
Entrez Gene ID
125228
Gene Name
family with sequence similarity 210 member A
Gene Aliases
C18orf19, HsT2329
UniGene
Hs.13034
Gene Chromosome Location
Chr.18: 13663347-13726592 on Build GRCh38
Transcripts
NM_001098801.1 NM_152352.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2884175 c.64A>G p.N22D Chr.18: 13682014-13682014 on Build GRCh38
4071401 c.42C>T p.R14R Chr.18: 13682036-13682036 on Build GRCh38
1522620 c.62A>G p.H21R Chr.18: 13682016-13682016 on Build GRCh38
7265336 c.20G>A p.R7Q Chr.18: 13682058-13682058 on Build GRCh38
6659419 c.63T>A p.H21Q Chr.18: 13682015-13682015 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs552796973 Chr.18: 13682038-13682038 on Build GRCh38
rs758549366 Chr.18: 13682030-13682030 on Build GRCh38
rs745561931 Chr.18: 13682058-13682058 on Build GRCh38
rs768981281 Chr.18: 13682074-13682074 on Build GRCh38
rs371076318 Chr.18: 13682059-13682059 on Build GRCh38
rs368263552 Chr.18: 13682123-13682123 on Build GRCh38
rs1009404284 Chr.18: 13682155-13682155 on Build GRCh38
rs1049672748 Chr.18: 13682048-13682048 on Build GRCh38
rs752482708 Chr.18: 13682041-13682041 on Build GRCh38
rs777985722 Chr.18: 13682198-13682198 on Build GRCh38
rs374211419 Chr.18: 13682099-13682099 on Build GRCh38
rs904849603 Chr.18: 13682089-13682089 on Build GRCh38
rs181249004 Chr.18: 13682093-13682093 on Build GRCh38
rs756838446 Chr.18: 13682056-13682056 on Build GRCh38
rs971238245 Chr.18: 13682160-13682160 on Build GRCh38
rs779671314 Chr.18: 13682065-13682065 on Build GRCh38
rs779670880 Chr.18: 13682036-13682037 on Build GRCh38
rs998684814 Chr.18: 13682169-13682169 on Build GRCh38
rs1012641850 Chr.18: 13682131-13682131 on Build GRCh38
rs762179618 Chr.18: 13682084-13682084 on Build GRCh38
rs568127171 Chr.18: 13682128-13682128 on Build GRCh38
rs774888776 Chr.18: 13682079-13682079 on Build GRCh38
rs186233628 Chr.18: 13682153-13682153 on Build GRCh38
rs1031542648 Chr.18: 13682187-13682187 on Build GRCh38
rs760768896 Chr.18: 13682037-13682037 on Build GRCh38
rs760965665 Chr.18: 13682096-13682096 on Build GRCh38
rs772224892 Chr.18: 13682086-13682086 on Build GRCh38
rs572460629 Chr.18: 13682051-13682051 on Build GRCh38
rs1022511531 Chr.18: 13682192-13682192 on Build GRCh38
rs754107601 Chr.18: 13682115-13682115 on Build GRCh38
rs200198941 Chr.18: 13682046-13682046 on Build GRCh38
rs188954860 Chr.18: 13682204-13682204 on Build GRCh38
rs749722333 Chr.18: 13682072-13682072 on Build GRCh38

View Full Product Details