Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 58716592-58716863 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GOT2
Entrez Gene ID
2806
Gene Name
glutamic-oxaloacetic transaminase 2
Gene Aliases
KAT4, KATIV, KYAT4, mitAAT
UniGene
Hs.599470
Gene Chromosome Location
Chr.16: 58707131-58734357 on Build GRCh38
Transcripts
NM_001286220.1 NM_002080.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6416664 c.797A>C p.E266A Chr.16: 58716719-58716719 on Build GRCh38
266603 c.719C>T p.A240V Chr.16: 58716797-58716797 on Build GRCh38
1128916 c.816C>T p.C272C Chr.16: 58716700-58716700 on Build GRCh38
3888721 c.795C>T p.I265I Chr.16: 58716721-58716721 on Build GRCh38
703987 c.835A>G p.K279E Chr.16: 58716681-58716681 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372350040 Chr.16: 58716833-58716833 on Build GRCh38
rs755980939 Chr.16: 58716621-58716621 on Build GRCh38
rs750151144 Chr.16: 58716787-58716787 on Build GRCh38
rs780177131 Chr.16: 58716629-58716629 on Build GRCh38
rs777550728 Chr.16: 58716825-58716825 on Build GRCh38
rs1003221168 Chr.16: 58716674-58716674 on Build GRCh38
rs745772197 Chr.16: 58716618-58716618 on Build GRCh38
rs747785290 Chr.16: 58716806-58716806 on Build GRCh38
rs764422535 Chr.16: 58716773-58716773 on Build GRCh38
rs572596398 Chr.16: 58716652-58716652 on Build GRCh38
rs368860781 Chr.16: 58716822-58716822 on Build GRCh38
rs928225766 Chr.16: 58716639-58716639 on Build GRCh38
rs138987040 Chr.16: 58716730-58716730 on Build GRCh38
rs752927520 Chr.16: 58716732-58716732 on Build GRCh38
rs141456453 Chr.16: 58716790-58716790 on Build GRCh38
rs552956269 Chr.16: 58716764-58716764 on Build GRCh38
rs780884521 Chr.16: 58716612-58716612 on Build GRCh38
rs374763381 Chr.16: 58716667-58716667 on Build GRCh38
rs760071011 Chr.16: 58716838-58716838 on Build GRCh38
rs150860349 Chr.16: 58716800-58716800 on Build GRCh38
rs758265732 Chr.16: 58716796-58716796 on Build GRCh38
rs117628895 Chr.16: 58716647-58716647 on Build GRCh38
rs894015371 Chr.16: 58716753-58716753 on Build GRCh38
rs756000870 Chr.16: 58716792-58716792 on Build GRCh38
rs138711511 Chr.16: 58716793-58716793 on Build GRCh38
rs772441242 Chr.16: 58716666-58716666 on Build GRCh38
rs777180671 Chr.16: 58716713-58716713 on Build GRCh38
rs763109950 Chr.16: 58716765-58716765 on Build GRCh38
rs777059898 Chr.16: 58716834-58716834 on Build GRCh38
rs368352559 Chr.16: 58716720-58716720 on Build GRCh38
rs768335492 Chr.16: 58716643-58716643 on Build GRCh38
rs867626398 Chr.16: 58716620-58716620 on Build GRCh38
rs766868890 Chr.16: 58716711-58716711 on Build GRCh38
rs754471368 Chr.16: 58716797-58716797 on Build GRCh38
rs1058192 Chr.16: 58716700-58716700 on Build GRCh38
rs866555564 Chr.16: 58716772-58716772 on Build GRCh38
rs748799019 Chr.16: 58716634-58716634 on Build GRCh38

View Full Product Details