Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 1092341-1092574 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C7orf50
Entrez Gene ID
84310
Gene Name
chromosome 7 open reading frame 50
Gene Aliases
YCR016W
UniGene
Hs.653258
Gene Chromosome Location
Chr.7: 978485-1138291 on Build GRCh38
Transcripts
NM_001134395.1 NM_001134396.1 NM_001318252.1 NM_032350.5 XM_011515581.2 XM_011515582.2 XM_011515583.2 XM_011515584.2 XM_017012720.1 XM_017012721.1 XM_017012722.1 NM_001039966.1 NM_001098201.1 NM_001505.2

Gene Symbol
GPER1
Entrez Gene ID
2852
Gene Name
G protein-coupled estrogen receptor 1
Gene Aliases
CEPR, CMKRL2, DRY12, FEG-1, GPCR-Br, GPER, GPR30, LERGU, LERGU2, LyGPR, mER
UniGene
Hs.20961
Gene Chromosome Location
Chr.7: 1086807-1093815 on Build GRCh38
Transcripts
NM_001134395.1 NM_001134396.1 NM_001318252.1 NM_032350.5 XM_011515581.2 XM_011515582.2 XM_011515583.2 XM_011515584.2 XM_017012720.1 XM_017012721.1 XM_017012722.1 NM_001039966.1 NM_001098201.1 NM_001505.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4687795 c.752G>A p.R251Q Chr.7: 1092480-1092480 on Build GRCh38
1084160 c.672C>T p.I224I Chr.7: 1092400-1092400 on Build GRCh38
4687794 c.751C>T p.R251W Chr.7: 1092479-1092479 on Build GRCh38
6248734 c.816G>T p.W272C Chr.7: 1092544-1092544 on Build GRCh38
6235654 c.632T>C p.V211A Chr.7: 1092360-1092360 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758236330 Chr.7: 1092499-1092499 on Build GRCh38
rs771510444 Chr.7: 1092401-1092401 on Build GRCh38
rs11977449 Chr.7: 1092439-1092439 on Build GRCh38
rs768379637 Chr.7: 1092394-1092394 on Build GRCh38
rs758076694 Chr.7: 1092452-1092452 on Build GRCh38
rs774757548 Chr.7: 1092479-1092479 on Build GRCh38
rs377670702 Chr.7: 1092481-1092481 on Build GRCh38
rs74331861 Chr.7: 1092514-1092514 on Build GRCh38
rs778619173 Chr.7: 1092400-1092400 on Build GRCh38
rs752930799 Chr.7: 1092370-1092370 on Build GRCh38
rs756699715 Chr.7: 1092505-1092505 on Build GRCh38
rs770631652 Chr.7: 1092541-1092541 on Build GRCh38
rs764231496 Chr.7: 1092428-1092428 on Build GRCh38
rs368234390 Chr.7: 1092418-1092418 on Build GRCh38
rs749090970 Chr.7: 1092389-1092389 on Build GRCh38
rs35069667 Chr.7: 1092471-1092471 on Build GRCh38
rs752906854 Chr.7: 1092422-1092422 on Build GRCh38
rs200314011 Chr.7: 1092556-1092556 on Build GRCh38
rs753789231 Chr.7: 1092444-1092444 on Build GRCh38
rs199736734 Chr.7: 1092380-1092380 on Build GRCh38
rs774415754 Chr.7: 1092460-1092460 on Build GRCh38
rs559603787 Chr.7: 1092532-1092532 on Build GRCh38
rs774706015 Chr.7: 1092528-1092528 on Build GRCh38
rs371278172 Chr.7: 1092464-1092464 on Build GRCh38
rs759628974 Chr.7: 1092363-1092363 on Build GRCh38
rs145985656 Chr.7: 1092459-1092459 on Build GRCh38
rs771448862 Chr.7: 1092410-1092410 on Build GRCh38
rs374724867 Chr.7: 1092489-1092489 on Build GRCh38
rs777183834 Chr.7: 1092463-1092463 on Build GRCh38
rs754230934 Chr.7: 1092362-1092362 on Build GRCh38
rs747835584 Chr.7: 1092480-1092480 on Build GRCh38
rs780572159 Chr.7: 1092512-1092512 on Build GRCh38
rs751714742 Chr.7: 1092436-1092436 on Build GRCh38
rs563630174 Chr.7: 1092488-1092488 on Build GRCh38
rs765060809 Chr.7: 1092547-1092547 on Build GRCh38
rs750319978 Chr.7: 1092382-1092382 on Build GRCh38
rs1053838604 Chr.7: 1092504-1092504 on Build GRCh38
rs747766901 Chr.7: 1092451-1092451 on Build GRCh38
rs139916786 Chr.7: 1092485-1092485 on Build GRCh38
rs772711029 Chr.7: 1092406-1092406 on Build GRCh38
rs952842968 Chr.7: 1092551-1092551 on Build GRCh38
rs777372818 Chr.7: 1092500-1092500 on Build GRCh38
rs778014567 Chr.7: 1092381-1092381 on Build GRCh38
rs3808352 Chr.7: 1092517-1092517 on Build GRCh38
rs767547288 Chr.7: 1092482-1092482 on Build GRCh38
rs762136910 Chr.7: 1092530-1092530 on Build GRCh38
rs1049225202 Chr.7: 1092419-1092419 on Build GRCh38
rs770131675 Chr.7: 1092465-1092465 on Build GRCh38
rs376978032 Chr.7: 1092536-1092536 on Build GRCh38
rs150646955 Chr.7: 1092387-1092387 on Build GRCh38
rs780113437 Chr.7: 1092443-1092443 on Build GRCh38
rs564861434 Chr.7: 1092516-1092516 on Build GRCh38
rs1025216784 Chr.7: 1092379-1092379 on Build GRCh38
rs958800961 Chr.7: 1092515-1092515 on Build GRCh38
rs746729641 Chr.7: 1092503-1092503 on Build GRCh38
rs756154497 Chr.7: 1092384-1092384 on Build GRCh38
rs202207715 Chr.7: 1092423-1092423 on Build GRCh38
rs777052431 Chr.7: 1092414-1092414 on Build GRCh38
rs760587019 Chr.7: 1092522-1092524 on Build GRCh38
rs766307448 Chr.7: 1092528-1092530 on Build GRCh38
rs763891231 Chr.7: 1092552-1092552 on Build GRCh38
rs543666614 Chr.7: 1092486-1092486 on Build GRCh38
rs369986460 Chr.7: 1092550-1092550 on Build GRCh38
rs367713059 Chr.7: 1092445-1092445 on Build GRCh38
rs146041906 Chr.7: 1092519-1092519 on Build GRCh38
rs773193769 Chr.7: 1092535-1092535 on Build GRCh38
rs139741139 Chr.7: 1092409-1092409 on Build GRCh38
rs148410177 Chr.7: 1092549-1092549 on Build GRCh38
rs765474183 Chr.7: 1092367-1092367 on Build GRCh38
rs775750584 Chr.7: 1092470-1092470 on Build GRCh38

View Full Product Details