Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 78274040-78274307 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJA4
Entrez Gene ID
55466
Gene Name
DnaJ heat shock protein family (Hsp40) member A4
Gene Aliases
MST104, MSTP104, PRO1472
UniGene
Hs.513053
Gene Chromosome Location
Chr.15: 78264145-78282196 on Build GRCh38
Transcripts
NM_001130182.1 NM_001130183.1 NM_018602.3 XM_005254535.3 XM_006720601.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7262304 c.422C>T p.P141L Chr.15: 78274281-78274281 on Build GRCh38
7096428 c.483C>T p.I161I Chr.15: 78274261-78274261 on Build GRCh38
7078704 c.419-1G>A p.? Chr.15: 78274196-78274196 on Build GRCh38
7078705 c.338-1G>A p.? Chr.15: 78274196-78274196 on Build GRCh38
965335 c.492G>T p.Q164H Chr.15: 78274270-78274270 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752111684 Chr.15: 78274247-78274247 on Build GRCh38
rs780832678 Chr.15: 78274132-78274132 on Build GRCh38
rs781343533 Chr.15: 78274162-78274162 on Build GRCh38
rs149661275 Chr.15: 78274284-78274284 on Build GRCh38
rs781633962 Chr.15: 78274252-78274252 on Build GRCh38
rs192360069 Chr.15: 78274161-78274161 on Build GRCh38
rs911884420 Chr.15: 78274108-78274108 on Build GRCh38
rs199951789 Chr.15: 78274061-78274062 on Build GRCh38
rs34666770 Chr.15: 78274233-78274234 on Build GRCh38
rs368915651 Chr.15: 78274218-78274218 on Build GRCh38
rs868055551 Chr.15: 78274281-78274281 on Build GRCh38
rs748997864 Chr.15: 78274168-78274168 on Build GRCh38
rs776251760 Chr.15: 78274224-78274224 on Build GRCh38
rs746263750 Chr.15: 78274256-78274256 on Build GRCh38
rs778772507 Chr.15: 78274276-78274276 on Build GRCh38
rs771854558 Chr.15: 78274182-78274182 on Build GRCh38
rs747457644 Chr.15: 78274159-78274159 on Build GRCh38
rs528353288 Chr.15: 78274065-78274065 on Build GRCh38
rs371681037 Chr.15: 78274219-78274219 on Build GRCh38
rs756513963 Chr.15: 78274262-78274262 on Build GRCh38
rs370037298 Chr.15: 78274146-78274146 on Build GRCh38
rs759776822 Chr.15: 78274228-78274228 on Build GRCh38
rs200635299 Chr.15: 78274233-78274233 on Build GRCh38
rs1036668202 Chr.15: 78274160-78274160 on Build GRCh38
rs757664742 Chr.15: 78274248-78274248 on Build GRCh38
rs1000996282 Chr.15: 78274110-78274110 on Build GRCh38
rs746165397 Chr.15: 78274191-78274192 on Build GRCh38
rs1032509870 Chr.15: 78274135-78274135 on Build GRCh38
rs897684172 Chr.15: 78274197-78274197 on Build GRCh38
rs747696932 Chr.15: 78274203-78274203 on Build GRCh38
rs771708077 Chr.15: 78274214-78274214 on Build GRCh38
rs768372244 Chr.15: 78274180-78274180 on Build GRCh38
rs770554000 Chr.15: 78274094-78274094 on Build GRCh38
rs771724336 Chr.15: 78274282-78274282 on Build GRCh38
rs774120961 Chr.15: 78274195-78274195 on Build GRCh38
rs140902481 Chr.15: 78274277-78274277 on Build GRCh38
rs758529211 Chr.15: 78274234-78274234 on Build GRCh38
rs61752772 Chr.15: 78274267-78274267 on Build GRCh38

View Full Product Details