Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 116717703-116717976 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C12orf49
Entrez Gene ID
79794
Gene Name
chromosome 12 open reading frame 49
Gene Aliases
-
UniGene
Hs.592011
Gene Chromosome Location
Chr.12: 116713320-116738070 on Build GRCh38
Transcripts
NM_024738.2 XM_005253937.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4866747 c.445-1G>T p.? Chr.12: 116717894-116717894 on Build GRCh38
3456624 c.477C>T p.P159P Chr.12: 116717861-116717861 on Build GRCh38
5493038 c.540G>A p.V180V Chr.12: 116717888-116717888 on Build GRCh38
6263577 c.543G>A p.Q181Q Chr.12: 116717885-116717885 on Build GRCh38
5493039 c.450G>A p.V150V Chr.12: 116717888-116717888 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755816626 Chr.12: 116717779-116717779 on Build GRCh38
rs35185105 Chr.12: 116717799-116717800 on Build GRCh38
rs780597452 Chr.12: 116717797-116717797 on Build GRCh38
rs749480171 Chr.12: 116717902-116717902 on Build GRCh38
rs777850901 Chr.12: 116717827-116717827 on Build GRCh38
rs199594769 Chr.12: 116717729-116717730 on Build GRCh38
rs373390806 Chr.12: 116717930-116717930 on Build GRCh38
rs780627404 Chr.12: 116717890-116717890 on Build GRCh38
rs758686738 Chr.12: 116717793-116717793 on Build GRCh38
rs750971832 Chr.12: 116717762-116717762 on Build GRCh38
rs770859795 Chr.12: 116717832-116717832 on Build GRCh38
rs371405515 Chr.12: 116717828-116717828 on Build GRCh38
rs143322155 Chr.12: 116717809-116717809 on Build GRCh38
rs771242241 Chr.12: 116717905-116717905 on Build GRCh38
rs377701665 Chr.12: 116717943-116717943 on Build GRCh38
rs201126216 Chr.12: 116717831-116717831 on Build GRCh38
rs755526029 Chr.12: 116717889-116717889 on Build GRCh38
rs376812526 Chr.12: 116717757-116717757 on Build GRCh38
rs939883851 Chr.12: 116717770-116717770 on Build GRCh38
rs960360185 Chr.12: 116717732-116717732 on Build GRCh38
rs745638178 Chr.12: 116717919-116717919 on Build GRCh38
rs112311710 Chr.12: 116717900-116717900 on Build GRCh38
rs373459286 Chr.12: 116717918-116717918 on Build GRCh38
rs772178056 Chr.12: 116717920-116717920 on Build GRCh38
rs759582903 Chr.12: 116717855-116717855 on Build GRCh38
rs908436892 Chr.12: 116717769-116717769 on Build GRCh38
rs768064953 Chr.12: 116717928-116717928 on Build GRCh38
rs752217115 Chr.12: 116717773-116717773 on Build GRCh38
rs546759950 Chr.12: 116717942-116717942 on Build GRCh38
rs200462423 Chr.12: 116717891-116717891 on Build GRCh38
rs976654441 Chr.12: 116717954-116717954 on Build GRCh38
rs750842035 Chr.12: 116717791-116717791 on Build GRCh38
rs752065284 Chr.12: 116717800-116717800 on Build GRCh38
rs147935392 Chr.12: 116717815-116717815 on Build GRCh38
rs371841873 Chr.12: 116717789-116717789 on Build GRCh38
rs777521532 Chr.12: 116717945-116717945 on Build GRCh38
rs753419585 Chr.12: 116717868-116717868 on Build GRCh38
rs146457169 Chr.12: 116717772-116717772 on Build GRCh38
rs774225642 Chr.12: 116717852-116717852 on Build GRCh38
rs776103501 Chr.12: 116717914-116717915 on Build GRCh38
rs928743561 Chr.12: 116717729-116717729 on Build GRCh38
rs760289623 Chr.12: 116717896-116717897 on Build GRCh38
rs891362057 Chr.12: 116717777-116717777 on Build GRCh38
rs751872632 Chr.12: 116717873-116717873 on Build GRCh38
rs375171430 Chr.12: 116717816-116717816 on Build GRCh38
rs766760609 Chr.12: 116717869-116717869 on Build GRCh38
rs1034963483 Chr.12: 116717880-116717880 on Build GRCh38
rs760290447 Chr.12: 116717923-116717923 on Build GRCh38
rs370448920 Chr.12: 116717921-116717921 on Build GRCh38
rs991544530 Chr.12: 116717748-116717748 on Build GRCh38
rs141818045 Chr.12: 116717857-116717857 on Build GRCh38
rs550643988 Chr.12: 116717856-116717856 on Build GRCh38
rs764846242 Chr.12: 116717932-116717932 on Build GRCh38
rs199517113 Chr.12: 116717859-116717859 on Build GRCh38
rs915905839 Chr.12: 116717751-116717751 on Build GRCh38
rs755438323 Chr.12: 116717803-116717803 on Build GRCh38
rs369230667 Chr.12: 116717929-116717929 on Build GRCh38
rs775152045 Chr.12: 116717832-116717832 on Build GRCh38
rs759493107 Chr.12: 116717768-116717768 on Build GRCh38
rs1053533878 Chr.12: 116717952-116717952 on Build GRCh38
rs934927537 Chr.12: 116717948-116717948 on Build GRCh38

View Full Product Details