Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 30417443-30417706 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MPPED2
Entrez Gene ID
744
Gene Name
metallophosphoesterase domain containing 2
Gene Aliases
239FB, C11orf8
UniGene
Hs.289795
Gene Chromosome Location
Chr.11: 30384493-30586713 on Build GRCh38
Transcripts
NM_001145399.1 NM_001584.2 XM_005253110.3 XM_005253111.2 XM_005253112.1 XM_011520346.1 XM_011520347.2 XM_017018231.1 XM_017018232.1 XM_017018233.1 XM_017018234.1 XM_017018235.1 XM_017018236.1 XM_017018237.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5764432 c.649C>G p.L217V Chr.11: 30417521-30417521 on Build GRCh38
5897801 c.611C>T p.P204L Chr.11: 30417559-30417559 on Build GRCh38
4502340 c.610C>T p.P204S Chr.11: 30417560-30417560 on Build GRCh38
687583 c.537-2A>T p.? Chr.11: 30417635-30417635 on Build GRCh38
6714884 c.643C>A p.P215T Chr.11: 30417527-30417527 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756763723 Chr.11: 30417678-30417678 on Build GRCh38
rs747862152 Chr.11: 30417563-30417563 on Build GRCh38
rs748936233 Chr.11: 30417677-30417677 on Build GRCh38
rs769345243 Chr.11: 30417471-30417471 on Build GRCh38
rs767302358 Chr.11: 30417658-30417658 on Build GRCh38
rs998551978 Chr.11: 30417602-30417602 on Build GRCh38
rs139073289 Chr.11: 30417540-30417540 on Build GRCh38
rs751402444 Chr.11: 30417542-30417542 on Build GRCh38
rs754808066 Chr.11: 30417545-30417545 on Build GRCh38
rs114983317 Chr.11: 30417657-30417657 on Build GRCh38
rs752430980 Chr.11: 30417661-30417661 on Build GRCh38
rs777355219 Chr.11: 30417576-30417576 on Build GRCh38
rs1012543735 Chr.11: 30417533-30417533 on Build GRCh38
rs939628011 Chr.11: 30417482-30417482 on Build GRCh38
rs764338218 Chr.11: 30417506-30417506 on Build GRCh38
rs149492180 Chr.11: 30417553-30417553 on Build GRCh38
rs375647995 Chr.11: 30417511-30417511 on Build GRCh38
rs961297692 Chr.11: 30417592-30417592 on Build GRCh38
rs377389874 Chr.11: 30417587-30417587 on Build GRCh38
rs767959397 Chr.11: 30417502-30417502 on Build GRCh38
rs764358251 Chr.11: 30417628-30417628 on Build GRCh38
rs571151311 Chr.11: 30417684-30417684 on Build GRCh38
rs1005311156 Chr.11: 30417550-30417550 on Build GRCh38
rs199958609 Chr.11: 30417505-30417505 on Build GRCh38
rs772620315 Chr.11: 30417600-30417600 on Build GRCh38
rs758372757 Chr.11: 30417529-30417529 on Build GRCh38
rs751501357 Chr.11: 30417649-30417649 on Build GRCh38
rs771284066 Chr.11: 30417481-30417481 on Build GRCh38
rs755785323 Chr.11: 30417664-30417664 on Build GRCh38
rs775994673 Chr.11: 30417626-30417626 on Build GRCh38
rs755671168 Chr.11: 30417470-30417471 on Build GRCh38
rs776799135 Chr.11: 30417638-30417638 on Build GRCh38
rs760986143 Chr.11: 30417627-30417627 on Build GRCh38
rs763519046 Chr.11: 30417478-30417478 on Build GRCh38
rs759775685 Chr.11: 30417499-30417499 on Build GRCh38
rs769367947 Chr.11: 30417574-30417574 on Build GRCh38
rs753014241 Chr.11: 30417503-30417503 on Build GRCh38
rs372235541 Chr.11: 30417683-30417683 on Build GRCh38
rs376718462 Chr.11: 30417471-30417472 on Build GRCh38
rs144774891 Chr.11: 30417597-30417597 on Build GRCh38
rs1015870417 Chr.11: 30417578-30417578 on Build GRCh38
rs772703095 Chr.11: 30417473-30417473 on Build GRCh38
rs986632302 Chr.11: 30417558-30417558 on Build GRCh38
rs953995725 Chr.11: 30417644-30417644 on Build GRCh38
rs371041959 Chr.11: 30417497-30417497 on Build GRCh38
rs777250603 Chr.11: 30417671-30417671 on Build GRCh38
rs774756838 Chr.11: 30417591-30417591 on Build GRCh38
rs780944866 Chr.11: 30417546-30417546 on Build GRCh38
rs765330644 Chr.11: 30417642-30417642 on Build GRCh38
rs748698110 Chr.11: 30417584-30417584 on Build GRCh38

View Full Product Details