Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 52703470-52703727 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LECT1
Entrez Gene ID
11061
Gene Name
leukocyte cell derived chemotaxin 1
Gene Aliases
BRICD3, CHM-I, CHM1, MYETS1
UniGene
Hs.421391
Gene Chromosome Location
Chr.13: 52703264-52739812 on Build GRCh38
Transcripts
NM_001011705.1 NM_007015.2 XM_011534897.2 XM_011534898.2 XM_011534899.2 XM_011534900.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5968235 c.914C>A p.P305Q Chr.13: 52703686-52703686 on Build GRCh38
5967728 c.973T>G p.W325G Chr.13: 52703627-52703627 on Build GRCh38
71423 c.914C>G p.P305R Chr.13: 52703686-52703686 on Build GRCh38
6042753 c.948C>A p.A316A Chr.13: 52703652-52703652 on Build GRCh38
6700361 c.931C>A p.Q311K Chr.13: 52703669-52703669 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775307491 Chr.13: 52703660-52703660 on Build GRCh38
rs774793940 Chr.13: 52703546-52703546 on Build GRCh38
rs961273691 Chr.13: 52703678-52703678 on Build GRCh38
rs149112371 Chr.13: 52703656-52703656 on Build GRCh38
rs770828040 Chr.13: 52703641-52703641 on Build GRCh38
rs569601718 Chr.13: 52703699-52703699 on Build GRCh38
rs1035986975 Chr.13: 52703636-52703636 on Build GRCh38
rs762304496 Chr.13: 52703547-52703547 on Build GRCh38
rs762565808 Chr.13: 52703665-52703665 on Build GRCh38
rs763230326 Chr.13: 52703689-52703701 on Build GRCh38
rs772880438 Chr.13: 52703630-52703631 on Build GRCh38
rs201954506 Chr.13: 52703605-52703605 on Build GRCh38
rs750923573 Chr.13: 52703552-52703552 on Build GRCh38
rs766810332 Chr.13: 52703680-52703680 on Build GRCh38
rs780788020 Chr.13: 52703649-52703649 on Build GRCh38
rs766477684 Chr.13: 52703576-52703576 on Build GRCh38
rs17541412 Chr.13: 52703537-52703537 on Build GRCh38
rs1052307 Chr.13: 52703577-52703577 on Build GRCh38
rs779135748 Chr.13: 52703615-52703615 on Build GRCh38
rs746958626 Chr.13: 52703638-52703638 on Build GRCh38
rs959966857 Chr.13: 52703646-52703646 on Build GRCh38
rs566918510 Chr.13: 52703522-52703522 on Build GRCh38
rs267603850 Chr.13: 52703706-52703706 on Build GRCh38
rs753976938 Chr.13: 52703614-52703614 on Build GRCh38
rs373527320 Chr.13: 52703705-52703705 on Build GRCh38
rs777466607 Chr.13: 52703637-52703637 on Build GRCh38
rs769580305 Chr.13: 52703544-52703544 on Build GRCh38
rs767864633 Chr.13: 52703549-52703549 on Build GRCh38
rs772453490 Chr.13: 52703670-52703670 on Build GRCh38
rs903403001 Chr.13: 52703543-52703543 on Build GRCh38
rs9536243 Chr.13: 52703504-52703504 on Build GRCh38
rs758188708 Chr.13: 52703633-52703633 on Build GRCh38
rs891271512 Chr.13: 52703499-52703499 on Build GRCh38
rs753874162 Chr.13: 52703697-52703697 on Build GRCh38
rs201037740 Chr.13: 52703676-52703676 on Build GRCh38
rs9536244 Chr.13: 52703511-52703511 on Build GRCh38
rs999518543 Chr.13: 52703550-52703550 on Build GRCh38
rs761127119 Chr.13: 52703556-52703556 on Build GRCh38
rs78575404 Chr.13: 52703655-52703655 on Build GRCh38
rs769459626 Chr.13: 52703612-52703613 on Build GRCh38
rs374765429 Chr.13: 52703493-52703493 on Build GRCh38
rs371116344 Chr.13: 52703616-52703616 on Build GRCh38
rs765348135 Chr.13: 52703702-52703702 on Build GRCh38
rs752884784 Chr.13: 52703703-52703703 on Build GRCh38

View Full Product Details