Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 136854751-136855004 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAMDC4
Entrez Gene ID
158056
Gene Name
MAM domain containing 4
Gene Aliases
AEGP
UniGene
Hs.376780
Gene Chromosome Location
Chr.9: 136852367-136860799 on Build GRCh38
Transcripts
NM_206920.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6362412 c.1073T>G p.L358R Chr.9: 136854986-136854986 on Build GRCh38
6033910 c.1032C>A p.F344L Chr.9: 136854945-136854945 on Build GRCh38
6706444 c.1003T>G p.S335A Chr.9: 136854830-136854830 on Build GRCh38
6033911 c.1032C>A p.F344L Chr.9: 136854945-136854945 on Build GRCh38
6706447 c.1021T>A p.S341T Chr.9: 136854848-136854848 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772344306 Chr.9: 136854878-136854878 on Build GRCh38
rs752827818 Chr.9: 136854961-136854961 on Build GRCh38
rs201155048 Chr.9: 136854902-136854902 on Build GRCh38
rs772381683 Chr.9: 136854806-136854806 on Build GRCh38
rs551209503 Chr.9: 136854932-136854932 on Build GRCh38
rs758902427 Chr.9: 136854834-136854834 on Build GRCh38
rs139787225 Chr.9: 136854828-136854828 on Build GRCh38
rs140616288 Chr.9: 136854870-136854870 on Build GRCh38
rs757513093 Chr.9: 136854981-136854981 on Build GRCh38
rs981838918 Chr.9: 136854963-136854963 on Build GRCh38
rs151315050 Chr.9: 136854823-136854823 on Build GRCh38
rs975490260 Chr.9: 136854874-136854874 on Build GRCh38
rs761449920 Chr.9: 136854917-136854917 on Build GRCh38
rs929866100 Chr.9: 136854801-136854801 on Build GRCh38
rs113437598 Chr.9: 136854838-136854838 on Build GRCh38
rs758483946 Chr.9: 136854962-136854962 on Build GRCh38
rs748352219 Chr.9: 136854877-136854877 on Build GRCh38
rs370418917 Chr.9: 136854867-136854867 on Build GRCh38
rs758224590 Chr.9: 136854769-136854769 on Build GRCh38
rs757207121 Chr.9: 136854859-136854859 on Build GRCh38
rs969152045 Chr.9: 136854947-136854947 on Build GRCh38
rs776962253 Chr.9: 136854885-136854885 on Build GRCh38
rs141677206 Chr.9: 136854777-136854777 on Build GRCh38
rs927775406 Chr.9: 136854983-136854983 on Build GRCh38
rs777204710 Chr.9: 136854832-136854832 on Build GRCh38
rs746329257 Chr.9: 136854985-136854985 on Build GRCh38
rs778955350 Chr.9: 136854945-136854945 on Build GRCh38
rs777901953 Chr.9: 136854965-136854965 on Build GRCh38
rs1045220189 Chr.9: 136854798-136854798 on Build GRCh38
rs916254907 Chr.9: 136854922-136854922 on Build GRCh38
rs377443370 Chr.9: 136854770-136854770 on Build GRCh38
rs534859465 Chr.9: 136854973-136854973 on Build GRCh38
rs372947541 Chr.9: 136854831-136854831 on Build GRCh38
rs1046195492 Chr.9: 136854778-136854778 on Build GRCh38
rs750282182 Chr.9: 136854933-136854933 on Build GRCh38
rs750381613 Chr.9: 136854865-136854865 on Build GRCh38
rs749388123 Chr.9: 136854784-136854784 on Build GRCh38
rs375996314 Chr.9: 136854864-136854864 on Build GRCh38
rs775180252 Chr.9: 136854891-136854891 on Build GRCh38
rs371603939 Chr.9: 136854888-136854888 on Build GRCh38
rs746920500 Chr.9: 136854773-136854773 on Build GRCh38
rs1044794699 Chr.9: 136854939-136854939 on Build GRCh38
rs761548657 Chr.9: 136854918-136854918 on Build GRCh38
rs780178024 Chr.9: 136854780-136854780 on Build GRCh38
rs544927460 Chr.9: 136854837-136854837 on Build GRCh38
rs370273159 Chr.9: 136854785-136854785 on Build GRCh38
rs763951533 Chr.9: 136854849-136854849 on Build GRCh38
rs144236396 Chr.9: 136854818-136854818 on Build GRCh38
rs748405373 Chr.9: 136854805-136854805 on Build GRCh38
rs771313012 Chr.9: 136854779-136854779 on Build GRCh38
rs368282227 Chr.9: 136854879-136854879 on Build GRCh38
rs150548116 Chr.9: 136854803-136854803 on Build GRCh38
rs139450438 Chr.9: 136854817-136854817 on Build GRCh38
rs144997410 Chr.9: 136854850-136854850 on Build GRCh38
rs769164513 Chr.9: 136854889-136854889 on Build GRCh38
rs756130110 Chr.9: 136854942-136854942 on Build GRCh38
rs747313651 Chr.9: 136854880-136854880 on Build GRCh38
rs762690787 Chr.9: 136854898-136854898 on Build GRCh38
rs114590925 Chr.9: 136854903-136854903 on Build GRCh38
rs747285315 Chr.9: 136854972-136854972 on Build GRCh38
rs998302586 Chr.9: 136854856-136854856 on Build GRCh38
rs114184364 Chr.9: 136854883-136854883 on Build GRCh38

View Full Product Details