Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 77857278-77857537 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAGT1
Entrez Gene ID
84061
Gene Name
magnesium transporter 1
Gene Aliases
IAP, MRX95, OST3B, PRO0756, XMEN, bA217H1.1
UniGene
Hs.323562
Gene Chromosome Location
Chr.X: 77826364-77895568 on Build GRCh38
Transcripts
NM_032121.5

SNP Detail

SNP ID Chromosome Location
rs368343097 Chr.X: 77857374-77857374 on Build GRCh38
rs782534818 Chr.X: 77857387-77857387 on Build GRCh38
rs782570442 Chr.X: 77857394-77857394 on Build GRCh38
rs782350149 Chr.X: 77857413-77857413 on Build GRCh38
rs782270890 Chr.X: 77857466-77857466 on Build GRCh38
rs782725051 Chr.X: 77857498-77857498 on Build GRCh38
rs181702408 Chr.X: 77857358-77857358 on Build GRCh38
rs782169515 Chr.X: 77857412-77857412 on Build GRCh38
rs782064334 Chr.X: 77857372-77857372 on Build GRCh38
rs782101769 Chr.X: 77857497-77857497 on Build GRCh38
rs782043620 Chr.X: 77857491-77857491 on Build GRCh38
rs781920327 Chr.X: 77857492-77857492 on Build GRCh38
rs1032635187 Chr.X: 77857439-77857439 on Build GRCh38
rs782379098 Chr.X: 77857484-77857484 on Build GRCh38
rs372142118 Chr.X: 77857386-77857386 on Build GRCh38
rs781805868 Chr.X: 77857503-77857503 on Build GRCh38
rs900993511 Chr.X: 77857326-77857326 on Build GRCh38
rs781980203 Chr.X: 77857499-77857499 on Build GRCh38
rs782796309 Chr.X: 77857500-77857500 on Build GRCh38
rs782603321 Chr.X: 77857437-77857437 on Build GRCh38
rs538518859 Chr.X: 77857317-77857317 on Build GRCh38
rs141774769 Chr.X: 77857377-77857377 on Build GRCh38
rs374161124 Chr.X: 77857366-77857366 on Build GRCh38
rs782632889 Chr.X: 77857385-77857385 on Build GRCh38

View Full Product Details