Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100577174-100577415 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRCH4
Entrez Gene ID
4034
Gene Name
leucine rich repeats and calponin homology domain containing 4
Gene Aliases
LRN, LRRN1, LRRN4, PP14183
UniGene
Hs.125742
Gene Chromosome Location
Chr.7: 100574011-100586188 on Build GRCh38
Transcripts
NM_001289934.1 NM_002319.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
743729 c.1284G>A p.P428P Chr.7: 100577284-100577284 on Build GRCh38
5715970 c.1221C>T p.T407T Chr.7: 100577347-100577347 on Build GRCh38
5710269 c.1294A>T p.S432C Chr.7: 100577274-100577274 on Build GRCh38
4650725 c.1253A>G p.Q418R Chr.7: 100577315-100577315 on Build GRCh38
6996253 c.1233G>A p.W411* Chr.7: 100577335-100577335 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769364186 Chr.7: 100577378-100577378 on Build GRCh38
rs372814020 Chr.7: 100577258-100577258 on Build GRCh38
rs767148562 Chr.7: 100577389-100577389 on Build GRCh38
rs770865237 Chr.7: 100577301-100577301 on Build GRCh38
rs769935166 Chr.7: 100577304-100577305 on Build GRCh38
rs748334008 Chr.7: 100577200-100577200 on Build GRCh38
rs768321694 Chr.7: 100577319-100577319 on Build GRCh38
rs371373030 Chr.7: 100577302-100577316 on Build GRCh38
rs772699164 Chr.7: 100577379-100577379 on Build GRCh38
rs761412341 Chr.7: 100577322-100577322 on Build GRCh38
rs775285384 Chr.7: 100577265-100577265 on Build GRCh38
rs756370433 Chr.7: 100577353-100577353 on Build GRCh38
rs775696132 Chr.7: 100577305-100577307 on Build GRCh38
rs751351053 Chr.7: 100577349-100577349 on Build GRCh38
rs369763673 Chr.7: 100577357-100577357 on Build GRCh38
rs759965571 Chr.7: 100577257-100577257 on Build GRCh38
rs757360489 Chr.7: 100577355-100577355 on Build GRCh38
rs765196008 Chr.7: 100577289-100577289 on Build GRCh38
rs762823669 Chr.7: 100577339-100577339 on Build GRCh38
rs763157567 Chr.7: 100577305-100577310 on Build GRCh38
rs753379286 Chr.7: 100577285-100577285 on Build GRCh38
rs747353792 Chr.7: 100577361-100577361 on Build GRCh38
rs140761835 Chr.7: 100577387-100577387 on Build GRCh38
rs749358816 Chr.7: 100577298-100577298 on Build GRCh38
rs371513153 Chr.7: 100577205-100577205 on Build GRCh38
rs150962857 Chr.7: 100577354-100577354 on Build GRCh38
rs749818546 Chr.7: 100577243-100577243 on Build GRCh38
rs777783218 Chr.7: 100577297-100577297 on Build GRCh38
rs779503954 Chr.7: 100577302-100577302 on Build GRCh38
rs776051139 Chr.7: 100577321-100577321 on Build GRCh38
rs142550780 Chr.7: 100577253-100577253 on Build GRCh38
rs369121637 Chr.7: 100577197-100577197 on Build GRCh38
rs200007538 Chr.7: 100577198-100577198 on Build GRCh38
rs768851548 Chr.7: 100577363-100577363 on Build GRCh38
rs773285298 Chr.7: 100577231-100577231 on Build GRCh38
rs756317139 Chr.7: 100577296-100577296 on Build GRCh38
rs747691986 Chr.7: 100577377-100577377 on Build GRCh38
rs933888653 Chr.7: 100577383-100577383 on Build GRCh38
rs746091769 Chr.7: 100577207-100577228 on Build GRCh38
rs760345848 Chr.7: 100577282-100577282 on Build GRCh38
rs368512611 Chr.7: 100577254-100577254 on Build GRCh38
rs767322495 Chr.7: 100577315-100577316 on Build GRCh38
rs1021681958 Chr.7: 100577350-100577350 on Build GRCh38
rs752933880 Chr.7: 100577294-100577294 on Build GRCh38
rs966970727 Chr.7: 100577326-100577326 on Build GRCh38
rs748921886 Chr.7: 100577388-100577388 on Build GRCh38
rs764615072 Chr.7: 100577327-100577327 on Build GRCh38
rs772577478 Chr.7: 100577316-100577316 on Build GRCh38
rs532401978 Chr.7: 100577213-100577213 on Build GRCh38
rs779761204 Chr.7: 100577291-100577291 on Build GRCh38
rs763750913 Chr.7: 100577284-100577284 on Build GRCh38
rs776381093 Chr.7: 100577368-100577368 on Build GRCh38
rs373208078 Chr.7: 100577358-100577358 on Build GRCh38
rs529847950 Chr.7: 100577287-100577287 on Build GRCh38
rs775980238 Chr.7: 100577317-100577317 on Build GRCh38
rs201757672 Chr.7: 100577328-100577328 on Build GRCh38
rs548446543 Chr.7: 100577347-100577347 on Build GRCh38
rs760466759 Chr.7: 100577318-100577318 on Build GRCh38
rs564345349 Chr.7: 100577206-100577206 on Build GRCh38
rs762945239 Chr.7: 100577385-100577385 on Build GRCh38
rs530626700 Chr.7: 100577360-100577360 on Build GRCh38

View Full Product Details