Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 105348313-105348556 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC44A1
Entrez Gene ID
23446
Gene Name
solute carrier family 44 member 1
Gene Aliases
CD92, CDW92, CHTL1, CTL1
UniGene
Hs.573495
Gene Chromosome Location
Chr.9: 105244608-105438504 on Build GRCh38
Transcripts
NM_001286730.1 NM_080546.4 XM_005251855.4 XM_006717027.3 XM_006717028.3 XM_006717029.2 XM_017014559.1 XM_017014560.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3413222 c.451A>G p.T151A Chr.9: 105348402-105348402 on Build GRCh38
7325099 c.407-1G>A p.? Chr.9: 105348357-105348357 on Build GRCh38
1103551 c.497C>T p.A166V Chr.9: 105348448-105348448 on Build GRCh38
285091 c.465T>G p.S155S Chr.9: 105348416-105348416 on Build GRCh38
5621851 c.449C>T p.T150I Chr.9: 105348400-105348400 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766212579 Chr.9: 105348492-105348492 on Build GRCh38
rs954615281 Chr.9: 105348398-105348398 on Build GRCh38
rs1049894898 Chr.9: 105348529-105348529 on Build GRCh38
rs747479186 Chr.9: 105348519-105348519 on Build GRCh38
rs878967123 Chr.9: 105348368-105348368 on Build GRCh38
rs768209372 Chr.9: 105348433-105348433 on Build GRCh38
rs575753512 Chr.9: 105348440-105348440 on Build GRCh38
rs760809347 Chr.9: 105348501-105348501 on Build GRCh38
rs920032974 Chr.9: 105348502-105348502 on Build GRCh38
rs199589545 Chr.9: 105348428-105348428 on Build GRCh38
rs932313004 Chr.9: 105348504-105348504 on Build GRCh38
rs762707611 Chr.9: 105348480-105348480 on Build GRCh38
rs74511615 Chr.9: 105348387-105348387 on Build GRCh38
rs779371576 Chr.9: 105348353-105348353 on Build GRCh38
rs769750343 Chr.9: 105348456-105348456 on Build GRCh38
rs374168300 Chr.9: 105348377-105348377 on Build GRCh38
rs776010289 Chr.9: 105348448-105348448 on Build GRCh38
rs780698255 Chr.9: 105348402-105348402 on Build GRCh38
rs746264700 Chr.9: 105348379-105348379 on Build GRCh38
rs752956207 Chr.9: 105348499-105348499 on Build GRCh38
rs772955444 Chr.9: 105348477-105348477 on Build GRCh38
rs200699902 Chr.9: 105348449-105348449 on Build GRCh38
rs879625998 Chr.9: 105348513-105348513 on Build GRCh38
rs773697834 Chr.9: 105348472-105348472 on Build GRCh38

View Full Product Details