Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 240130942-240131203 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COPS9
Entrez Gene ID
150678
Gene Name
COP9 signalosome subunit 9
Gene Aliases
CSNAP, MYEOV2
UniGene
Hs.293884
Gene Chromosome Location
Chr.2: 240126548-240136802 on Build GRCh38
Transcripts
NM_001163424.1 NM_138336.1 XM_017003411.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3839036 c.230-1G>C p.? Chr.2: 240131089-240131089 on Build GRCh38
6010745 c.263+5G>C p.? Chr.2: 240131050-240131050 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs759610174 Chr.2: 240131151-240131151 on Build GRCh38
rs112556280 Chr.2: 240130983-240130983 on Build GRCh38
rs755047625 Chr.2: 240131031-240131031 on Build GRCh38
rs973200484 Chr.2: 240131030-240131030 on Build GRCh38
rs759140163 Chr.2: 240131071-240131071 on Build GRCh38
rs767811368 Chr.2: 240131012-240131012 on Build GRCh38
rs758591766 Chr.2: 240131129-240131129 on Build GRCh38
rs769280248 Chr.2: 240131077-240131077 on Build GRCh38
rs757765838 Chr.2: 240131006-240131006 on Build GRCh38
rs777831667 Chr.2: 240131134-240131134 on Build GRCh38
rs1041738157 Chr.2: 240131165-240131165 on Build GRCh38
rs750965966 Chr.2: 240131104-240131104 on Build GRCh38
rs1008402286 Chr.2: 240130973-240130973 on Build GRCh38
rs780288468 Chr.2: 240131024-240131024 on Build GRCh38
rs778865933 Chr.2: 240131032-240131032 on Build GRCh38
rs771251981 Chr.2: 240131061-240131063 on Build GRCh38
rs752064588 Chr.2: 240131005-240131005 on Build GRCh38
rs775454822 Chr.2: 240131093-240131093 on Build GRCh38
rs562828218 Chr.2: 240131141-240131141 on Build GRCh38
rs200187046 Chr.2: 240131023-240131023 on Build GRCh38
rs774994682 Chr.2: 240131084-240131084 on Build GRCh38
rs377520178 Chr.2: 240131111-240131111 on Build GRCh38
rs755171414 Chr.2: 240131124-240131124 on Build GRCh38
rs903437931 Chr.2: 240131170-240131170 on Build GRCh38
rs759985670 Chr.2: 240131121-240131122 on Build GRCh38
rs776299995 Chr.2: 240131064-240131064 on Build GRCh38
rs369805109 Chr.2: 240131120-240131120 on Build GRCh38
rs980223834 Chr.2: 240131022-240131022 on Build GRCh38
rs200787829 Chr.2: 240131126-240131126 on Build GRCh38
rs774424389 Chr.2: 240131110-240131111 on Build GRCh38
rs927143142 Chr.2: 240131054-240131054 on Build GRCh38
rs772126954 Chr.2: 240131049-240131049 on Build GRCh38
rs750769707 Chr.2: 240131013-240131013 on Build GRCh38
rs940639002 Chr.2: 240131121-240131121 on Build GRCh38
rs1021664423 Chr.2: 240130996-240130996 on Build GRCh38
rs746769509 Chr.2: 240131052-240131052 on Build GRCh38
rs1050563812 Chr.2: 240131168-240131168 on Build GRCh38
rs748044636 Chr.2: 240131041-240131041 on Build GRCh38
rs766608650 Chr.2: 240130988-240130988 on Build GRCh38
rs752824250 Chr.2: 240131128-240131128 on Build GRCh38
rs761003068 Chr.2: 240131110-240131110 on Build GRCh38
rs571094024 Chr.2: 240131100-240131100 on Build GRCh38
rs981798386 Chr.2: 240130991-240130991 on Build GRCh38
rs756462534 Chr.2: 240130989-240130989 on Build GRCh38
rs770681255 Chr.2: 240131056-240131056 on Build GRCh38
rs750494334 Chr.2: 240130990-240130990 on Build GRCh38
rs956335254 Chr.2: 240130987-240130987 on Build GRCh38
rs773350804 Chr.2: 240131051-240131051 on Build GRCh38

View Full Product Details