Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 102606472-102606743 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MMP20
Entrez Gene ID
9313
Gene Name
matrix metallopeptidase 20
Gene Aliases
AI2A2, MMP-20
UniGene
Hs.591946
Gene Chromosome Location
Chr.11: 102576835-102625332 on Build GRCh38
Transcripts
NM_004771.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
685786 c.848C>A p.P283H Chr.11: 102606640-102606640 on Build GRCh38
3868278 c.857C>T p.P286L Chr.11: 102606631-102606631 on Build GRCh38
3397357 c.818G>A p.R273Q Chr.11: 102606670-102606670 on Build GRCh38
540616 c.833G>A p.G278E Chr.11: 102606655-102606655 on Build GRCh38
4835794 c.899C>G p.S300* Chr.11: 102606589-102606589 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757896780 Chr.11: 102606619-102606619 on Build GRCh38
rs1784423 Chr.11: 102606664-102606664 on Build GRCh38
rs538994781 Chr.11: 102606671-102606671 on Build GRCh38
rs769574714 Chr.11: 102606706-102606706 on Build GRCh38
rs749306471 Chr.11: 102606639-102606639 on Build GRCh38
rs200012206 Chr.11: 102606520-102606520 on Build GRCh38
rs755285702 Chr.11: 102606517-102606517 on Build GRCh38
rs780874817 Chr.11: 102606696-102606696 on Build GRCh38
rs116445633 Chr.11: 102606495-102606495 on Build GRCh38
rs146876571 Chr.11: 102606605-102606605 on Build GRCh38
rs1784424 Chr.11: 102606646-102606646 on Build GRCh38
rs779665059 Chr.11: 102606543-102606543 on Build GRCh38
rs544247299 Chr.11: 102606513-102606513 on Build GRCh38
rs747943008 Chr.11: 102606634-102606634 on Build GRCh38
rs763105283 Chr.11: 102606577-102606577 on Build GRCh38
rs35740710 Chr.11: 102606528-102606528 on Build GRCh38
rs770947538 Chr.11: 102606719-102606719 on Build GRCh38
rs764556174 Chr.11: 102606615-102606615 on Build GRCh38
rs754609325 Chr.11: 102606630-102606630 on Build GRCh38
rs768072719 Chr.11: 102606555-102606555 on Build GRCh38
rs746739126 Chr.11: 102606555-102606555 on Build GRCh38
rs371882715 Chr.11: 102606528-102606528 on Build GRCh38
rs757700559 Chr.11: 102606675-102606675 on Build GRCh38
rs757946027 Chr.11: 102606538-102606538 on Build GRCh38
rs763154703 Chr.11: 102606514-102606514 on Build GRCh38
rs746511120 Chr.11: 102606622-102606622 on Build GRCh38
rs762804684 Chr.11: 102606717-102606717 on Build GRCh38
rs141875245 Chr.11: 102606670-102606670 on Build GRCh38
rs796378193 Chr.11: 102606636-102606636 on Build GRCh38
rs761739576 Chr.11: 102606506-102606506 on Build GRCh38
rs778278609 Chr.11: 102606535-102606535 on Build GRCh38
rs145529899 Chr.11: 102606561-102606561 on Build GRCh38
rs759736667 Chr.11: 102606583-102606583 on Build GRCh38
rs751881766 Chr.11: 102606516-102606516 on Build GRCh38
rs752458039 Chr.11: 102606702-102606702 on Build GRCh38
rs767651260 Chr.11: 102606589-102606589 on Build GRCh38
rs776429623 Chr.11: 102606563-102606563 on Build GRCh38
rs754485319 Chr.11: 102606691-102606691 on Build GRCh38
rs780696972 Chr.11: 102606500-102606500 on Build GRCh38
rs148818720 Chr.11: 102606578-102606578 on Build GRCh38
rs17098826 Chr.11: 102606522-102606522 on Build GRCh38
rs747065790 Chr.11: 102606617-102606617 on Build GRCh38
rs145296288 Chr.11: 102606579-102606579 on Build GRCh38
rs375255666 Chr.11: 102606711-102606711 on Build GRCh38
rs773320444 Chr.11: 102606572-102606572 on Build GRCh38
rs755971350 Chr.11: 102606710-102606710 on Build GRCh38
rs187593820 Chr.11: 102606718-102606718 on Build GRCh38
rs774431820 Chr.11: 102606656-102606656 on Build GRCh38
rs766676232 Chr.11: 102606515-102606515 on Build GRCh38
rs751040687 Chr.11: 102606690-102606690 on Build GRCh38
rs769812507 Chr.11: 102606566-102606566 on Build GRCh38
rs17098831 Chr.11: 102606618-102606618 on Build GRCh38
rs746297837 Chr.11: 102606505-102606505 on Build GRCh38
rs779468348 Chr.11: 102606620-102606620 on Build GRCh38
rs369545672 Chr.11: 102606510-102606510 on Build GRCh38
rs201174058 Chr.11: 102606663-102606663 on Build GRCh38
rs756590286 Chr.11: 102606534-102606534 on Build GRCh38
rs765685299 Chr.11: 102606676-102606676 on Build GRCh38
rs141603134 Chr.11: 102606606-102606606 on Build GRCh38
rs760362822 Chr.11: 102606498-102606498 on Build GRCh38
rs759559840 Chr.11: 102606657-102606657 on Build GRCh38
rs768882792 Chr.11: 102606633-102606633 on Build GRCh38
rs182862570 Chr.11: 102606635-102606635 on Build GRCh38
rs149696445 Chr.11: 102606536-102606536 on Build GRCh38

View Full Product Details