Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 223957438-223957711 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MRPL44
Entrez Gene ID
65080
Gene Name
mitochondrial ribosomal protein L44
Gene Aliases
COXPD16, L44MT, MRP-L44
UniGene
Hs.203559
Gene Chromosome Location
Chr.2: 223943224-223967714 on Build GRCh38
Transcripts
NM_022915.3 XM_011511668.2 XM_017004730.1 XM_017004731.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5816281 c.41G>T p.R14L Chr.2: 223957513-223957513 on Build GRCh38
312938 c.135A>T p.L45F Chr.2: 223957607-223957607 on Build GRCh38
7098242 c.101G>A p.G34E Chr.2: 223957573-223957573 on Build GRCh38
4525115 c.130G>A p.E44K Chr.2: 223957602-223957602 on Build GRCh38
269620 c.38A>G p.H13R Chr.2: 223957510-223957510 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756922509 Chr.2: 223957579-223957579 on Build GRCh38
rs887524457 Chr.2: 223957675-223957675 on Build GRCh38
rs760943515 Chr.2: 223957540-223957540 on Build GRCh38
rs774616345 Chr.2: 223957527-223957527 on Build GRCh38
rs776802929 Chr.2: 223957562-223957562 on Build GRCh38
rs376994749 Chr.2: 223957639-223957639 on Build GRCh38
rs370632812 Chr.2: 223957480-223957480 on Build GRCh38
rs936019116 Chr.2: 223957556-223957556 on Build GRCh38
rs769887825 Chr.2: 223957591-223957591 on Build GRCh38
rs754177649 Chr.2: 223957546-223957546 on Build GRCh38
rs368212494 Chr.2: 223957493-223957493 on Build GRCh38
rs766910797 Chr.2: 223957491-223957491 on Build GRCh38
rs776375666 Chr.2: 223957472-223957472 on Build GRCh38
rs764338057 Chr.2: 223957478-223957478 on Build GRCh38
rs760075011 Chr.2: 223957549-223957549 on Build GRCh38
rs765747156 Chr.2: 223957646-223957646 on Build GRCh38
rs889706283 Chr.2: 223957575-223957575 on Build GRCh38
rs765692067 Chr.2: 223957550-223957550 on Build GRCh38
rs780095103 Chr.2: 223957594-223957594 on Build GRCh38
rs975710095 Chr.2: 223957633-223957633 on Build GRCh38
rs140294613 Chr.2: 223957506-223957506 on Build GRCh38
rs138134784 Chr.2: 223957620-223957620 on Build GRCh38
rs757845862 Chr.2: 223957562-223957562 on Build GRCh38
rs751060990 Chr.2: 223957682-223957682 on Build GRCh38
rs141306066 Chr.2: 223957577-223957577 on Build GRCh38
rs752226588 Chr.2: 223957554-223957554 on Build GRCh38
rs761948864 Chr.2: 223957533-223957533 on Build GRCh38
rs534596036 Chr.2: 223957510-223957510 on Build GRCh38
rs753136403 Chr.2: 223957650-223957650 on Build GRCh38
rs746740405 Chr.2: 223957463-223957463 on Build GRCh38
rs756372023 Chr.2: 223957490-223957490 on Build GRCh38
rs769836108 Chr.2: 223957683-223957683 on Build GRCh38
rs759844256 Chr.2: 223957644-223957644 on Build GRCh38
rs767807980 Chr.2: 223957534-223957534 on Build GRCh38
rs370753182 Chr.2: 223957568-223957568 on Build GRCh38
rs373144312 Chr.2: 223957686-223957686 on Build GRCh38
rs551735456 Chr.2: 223957497-223957497 on Build GRCh38
rs763131858 Chr.2: 223957473-223957473 on Build GRCh38
rs773236665 Chr.2: 223957535-223957535 on Build GRCh38
rs201475876 Chr.2: 223957502-223957502 on Build GRCh38
rs760852652 Chr.2: 223957608-223957608 on Build GRCh38
rs779436306 Chr.2: 223957523-223957523 on Build GRCh38
rs374474562 Chr.2: 223957586-223957586 on Build GRCh38
rs750835773 Chr.2: 223957489-223957489 on Build GRCh38
rs143105143 Chr.2: 223957581-223957581 on Build GRCh38
rs769834883 Chr.2: 223957521-223957521 on Build GRCh38
rs747624821 Chr.2: 223957503-223957503 on Build GRCh38
rs113422842 Chr.2: 223957512-223957512 on Build GRCh38
rs748238947 Chr.2: 223957601-223957601 on Build GRCh38
rs772107545 Chr.2: 223957603-223957603 on Build GRCh38
rs375576699 Chr.2: 223957674-223957674 on Build GRCh38
rs148281067 Chr.2: 223957519-223957519 on Build GRCh38
rs370122608 Chr.2: 223957660-223957660 on Build GRCh38
rs555765290 Chr.2: 223957613-223957613 on Build GRCh38
rs1050354867 Chr.2: 223957564-223957564 on Build GRCh38
rs766708259 Chr.2: 223957541-223957541 on Build GRCh38
rs770566525 Chr.2: 223957515-223957515 on Build GRCh38
rs556415712 Chr.2: 223957649-223957649 on Build GRCh38
rs199583883 Chr.2: 223957468-223957468 on Build GRCh38
rs748269428 Chr.2: 223957632-223957634 on Build GRCh38
rs368889641 Chr.2: 223957488-223957488 on Build GRCh38
rs375126881 Chr.2: 223957485-223957485 on Build GRCh38

View Full Product Details