Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 73000358-73000591 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SFXN5
Entrez Gene ID
94097
Gene Name
sideroflexin 5
Gene Aliases
BBG-TCC
UniGene
Hs.368171
Gene Chromosome Location
Chr.2: 72942036-73072234 on Build GRCh38
Transcripts
NM_144579.2 XM_005264648.1 XM_006712147.1 XM_011533186.2 XM_011533187.2 XM_011533188.2 XM_011533189.1 XM_011533190.2 XM_011533192.2 XM_011533194.2 XM_011533195.2 XM_011533196.2 XM_011533197.1 XM_017005340.1 XM_017005341.1 XM_017005342.1 XM_017005343.1 XM_017005344.1 XM_017005346.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1225490 c.455G>A p.R152H Chr.2: 73000444-73000444 on Build GRCh38
4964087 c.446A>T p.Y149F Chr.2: 73000453-73000453 on Build GRCh38
4964158 c.445T>A p.Y149N Chr.2: 73000454-73000454 on Build GRCh38
4646696 c.432T>C p.N144N Chr.2: 73000467-73000467 on Build GRCh38
4650611 c.461C>T p.A154V Chr.2: 73000438-73000438 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1054679015 Chr.2: 73000422-73000422 on Build GRCh38
rs143169685 Chr.2: 73000417-73000417 on Build GRCh38
rs865832853 Chr.2: 73000571-73000571 on Build GRCh38
rs112844917 Chr.2: 73000406-73000406 on Build GRCh38
rs951619227 Chr.2: 73000563-73000563 on Build GRCh38
rs772981750 Chr.2: 73000458-73000458 on Build GRCh38
rs35766484 Chr.2: 73000573-73000574 on Build GRCh38
rs201307864 Chr.2: 73000525-73000525 on Build GRCh38
rs778116077 Chr.2: 73000399-73000399 on Build GRCh38
rs573966519 Chr.2: 73000383-73000383 on Build GRCh38
rs188234508 Chr.2: 73000567-73000567 on Build GRCh38
rs748516012 Chr.2: 73000462-73000462 on Build GRCh38
rs1039856549 Chr.2: 73000526-73000530 on Build GRCh38
rs866397176 Chr.2: 73000444-73000444 on Build GRCh38
rs758638981 Chr.2: 73000392-73000392 on Build GRCh38
rs146657142 Chr.2: 73000449-73000449 on Build GRCh38
rs369326386 Chr.2: 73000438-73000438 on Build GRCh38
rs906505474 Chr.2: 73000467-73000467 on Build GRCh38
rs770429927 Chr.2: 73000470-73000470 on Build GRCh38
rs1003496806 Chr.2: 73000493-73000493 on Build GRCh38
rs779834270 Chr.2: 73000426-73000426 on Build GRCh38
rs760070416 Chr.2: 73000463-73000463 on Build GRCh38
rs368259333 Chr.2: 73000389-73000389 on Build GRCh38
rs894328858 Chr.2: 73000432-73000432 on Build GRCh38
rs1053929885 Chr.2: 73000556-73000556 on Build GRCh38
rs778611888 Chr.2: 73000441-73000441 on Build GRCh38
rs747949729 Chr.2: 73000445-73000445 on Build GRCh38
rs372579670 Chr.2: 73000531-73000531 on Build GRCh38
rs936809295 Chr.2: 73000554-73000554 on Build GRCh38
rs776038860 Chr.2: 73000503-73000503 on Build GRCh38
rs749198727 Chr.2: 73000437-73000437 on Build GRCh38
rs537808466 Chr.2: 73000386-73000386 on Build GRCh38
rs36093719 Chr.2: 73000534-73000535 on Build GRCh38
rs868611481 Chr.2: 73000485-73000485 on Build GRCh38

View Full Product Details