Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 7921676-7921937 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNAPC2
Entrez Gene ID
6618
Gene Name
small nuclear RNA activating complex polypeptide 2
Gene Aliases
PTFDELTA, SNAP45
UniGene
Hs.631860
Gene Chromosome Location
Chr.19: 7920309-7923251 on Build GRCh38
Transcripts
NM_003083.3 NR_030717.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5857128 c.372+1G>A p.? Chr.19: 7921774-7921774 on Build GRCh38
5772528 c.326A>T p.K109M Chr.19: 7921727-7921727 on Build GRCh38
6785923 c.312G>A p.T104T Chr.19: 7921713-7921713 on Build GRCh38
3893923 c.335G>A p.G112E Chr.19: 7921736-7921736 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs985824075 Chr.19: 7921778-7921778 on Build GRCh38
rs146799171 Chr.19: 7921738-7921738 on Build GRCh38
rs765710059 Chr.19: 7921744-7921744 on Build GRCh38
rs141299337 Chr.19: 7921716-7921716 on Build GRCh38
rs932907651 Chr.19: 7921732-7921732 on Build GRCh38
rs201639553 Chr.19: 7921766-7921766 on Build GRCh38
rs751378362 Chr.19: 7921874-7921874 on Build GRCh38
rs147722010 Chr.19: 7921900-7921900 on Build GRCh38
rs1055014035 Chr.19: 7921713-7921713 on Build GRCh38
rs372400847 Chr.19: 7921797-7921797 on Build GRCh38
rs777028828 Chr.19: 7921807-7921807 on Build GRCh38
rs2303698 Chr.19: 7921712-7921712 on Build GRCh38
rs1002534650 Chr.19: 7921833-7921833 on Build GRCh38
rs562827802 Chr.19: 7921780-7921780 on Build GRCh38
rs780299508 Chr.19: 7921799-7921799 on Build GRCh38
rs754648462 Chr.19: 7921763-7921763 on Build GRCh38
rs757062109 Chr.19: 7921888-7921888 on Build GRCh38
rs937917282 Chr.19: 7921835-7921835 on Build GRCh38
rs770332232 Chr.19: 7921811-7921811 on Build GRCh38
rs929172721 Chr.19: 7921794-7921794 on Build GRCh38
rs780862302 Chr.19: 7921707-7921707 on Build GRCh38
rs776886834 Chr.19: 7921741-7921741 on Build GRCh38
rs475002 Chr.19: 7921753-7921753 on Build GRCh38
rs760990578 Chr.19: 7921734-7921734 on Build GRCh38
rs747935820 Chr.19: 7921769-7921769 on Build GRCh38
rs7250749 Chr.19: 7921800-7921800 on Build GRCh38
rs754705554 Chr.19: 7921704-7921704 on Build GRCh38
rs530686592 Chr.19: 7921904-7921904 on Build GRCh38
rs1042465136 Chr.19: 7921884-7921884 on Build GRCh38
rs944456466 Chr.19: 7921773-7921773 on Build GRCh38
rs751154824 Chr.19: 7921755-7921755 on Build GRCh38
rs746805278 Chr.19: 7921699-7921699 on Build GRCh38
rs779742609 Chr.19: 7921694-7921694 on Build GRCh38
rs777636439 Chr.19: 7921785-7921785 on Build GRCh38
rs762752619 Chr.19: 7921850-7921850 on Build GRCh38
rs902573805 Chr.19: 7921894-7921894 on Build GRCh38
rs768798309 Chr.19: 7921804-7921804 on Build GRCh38
rs375557441 Chr.19: 7921779-7921779 on Build GRCh38
rs192927760 Chr.19: 7921843-7921843 on Build GRCh38
rs762347980 Chr.19: 7921743-7921743 on Build GRCh38
rs190413008 Chr.19: 7921715-7921715 on Build GRCh38
rs769651513 Chr.19: 7921710-7921710 on Build GRCh38
rs369842802 Chr.19: 7921789-7921789 on Build GRCh38
rs893790829 Chr.19: 7921880-7921880 on Build GRCh38
rs946832081 Chr.19: 7921881-7921881 on Build GRCh38
rs35980787 Chr.19: 7921833-7921834 on Build GRCh38
rs78470363 Chr.19: 7921809-7921809 on Build GRCh38
rs749938149 Chr.19: 7921750-7921750 on Build GRCh38
rs1021075533 Chr.19: 7921902-7921902 on Build GRCh38
rs143542798 Chr.19: 7921727-7921727 on Build GRCh38
rs998369323 Chr.19: 7921896-7921896 on Build GRCh38
rs757974392 Chr.19: 7921752-7921752 on Build GRCh38
rs772418257 Chr.19: 7921719-7921719 on Build GRCh38

View Full Product Details