Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 44803305-44803552 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLK3
Entrez Gene ID
1263
Gene Name
polo like kinase 3
Gene Aliases
CNK, FNK, PLK-3, PRK
UniGene
Hs.632415
Gene Chromosome Location
Chr.1: 44799952-44805995 on Build GRCh38
Transcripts
NM_004073.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1343002 c.889delC p.N299fs*35 Chr.1: 44803325-44803325 on Build GRCh38
6750204 c.1065G>T p.K355N Chr.1: 44803384-44803384 on Build GRCh38
4715798 c.948G>T p.K316N Chr.1: 44803384-44803384 on Build GRCh38
6750216 c.1006delC p.N338fs*35 Chr.1: 44803325-44803325 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1007319067 Chr.1: 44803435-44803435 on Build GRCh38
rs141933667 Chr.1: 44803391-44803391 on Build GRCh38
rs72674040 Chr.1: 44803474-44803474 on Build GRCh38
rs771413266 Chr.1: 44803405-44803405 on Build GRCh38
rs535901302 Chr.1: 44803425-44803425 on Build GRCh38
rs545003745 Chr.1: 44803512-44803512 on Build GRCh38
rs375317245 Chr.1: 44803365-44803365 on Build GRCh38
rs967048044 Chr.1: 44803497-44803497 on Build GRCh38
rs1032107383 Chr.1: 44803438-44803438 on Build GRCh38
rs143812663 Chr.1: 44803333-44803333 on Build GRCh38
rs866108502 Chr.1: 44803380-44803380 on Build GRCh38
rs555840434 Chr.1: 44803431-44803431 on Build GRCh38
rs768091554 Chr.1: 44803342-44803342 on Build GRCh38
rs981937897 Chr.1: 44803498-44803498 on Build GRCh38
rs200867561 Chr.1: 44803370-44803370 on Build GRCh38
rs774992582 Chr.1: 44803422-44803422 on Build GRCh38
rs753895242 Chr.1: 44803442-44803442 on Build GRCh38
rs112574988 Chr.1: 44803325-44803325 on Build GRCh38
rs764101227 Chr.1: 44803437-44803437 on Build GRCh38
rs745599478 Chr.1: 44803328-44803328 on Build GRCh38
rs879080890 Chr.1: 44803485-44803485 on Build GRCh38
rs1010604969 Chr.1: 44803452-44803452 on Build GRCh38
rs772542386 Chr.1: 44803430-44803430 on Build GRCh38
rs779744523 Chr.1: 44803335-44803335 on Build GRCh38
rs367908756 Chr.1: 44803436-44803436 on Build GRCh38
rs746645815 Chr.1: 44803336-44803336 on Build GRCh38

View Full Product Details