Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 37385968-37386227 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SRC
Entrez Gene ID
6714
Gene Name
SRC proto-oncogene, non-receptor tyrosine kinase
Gene Aliases
ASV, SRC1, THC6, c-SRC, p60-Src
UniGene
Hs.195659
Gene Chromosome Location
Chr.20: 37344685-37405432 on Build GRCh38
Transcripts
NM_005417.4 NM_198291.2 XM_011529013.2 XM_017028024.1 XM_017028025.1 XM_017028026.1 XM_017028027.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6453825 c.350C>T p.T117I Chr.20: 37386174-37386174 on Build GRCh38
1026507 c.272C>T p.A91V Chr.20: 37386096-37386096 on Build GRCh38
6790026 c.285T>C p.Y95Y Chr.20: 37386109-37386109 on Build GRCh38
5589037 c.333C>T p.L111L Chr.20: 37386157-37386157 on Build GRCh38
4729923 c.330G>A p.R110R Chr.20: 37386154-37386154 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372817033 Chr.20: 37386063-37386063 on Build GRCh38
rs746328956 Chr.20: 37386033-37386043 on Build GRCh38
rs772219397 Chr.20: 37386152-37386152 on Build GRCh38
rs869131587 Chr.20: 37386042-37386042 on Build GRCh38
rs199914043 Chr.20: 37386198-37386198 on Build GRCh38
rs755760984 Chr.20: 37386080-37386080 on Build GRCh38
rs149493327 Chr.20: 37386200-37386200 on Build GRCh38
rs777586605 Chr.20: 37386081-37386081 on Build GRCh38
rs763506536 Chr.20: 37386190-37386190 on Build GRCh38
rs377182327 Chr.20: 37386155-37386155 on Build GRCh38
rs752406826 Chr.20: 37386072-37386072 on Build GRCh38
rs562849105 Chr.20: 37386169-37386169 on Build GRCh38
rs778675662 Chr.20: 37386118-37386118 on Build GRCh38
rs967033337 Chr.20: 37386008-37386008 on Build GRCh38
rs769864399 Chr.20: 37386088-37386089 on Build GRCh38
rs747298865 Chr.20: 37386153-37386153 on Build GRCh38
rs765782972 Chr.20: 37386181-37386181 on Build GRCh38
rs550612252 Chr.20: 37386121-37386121 on Build GRCh38
rs528748634 Chr.20: 37386023-37386023 on Build GRCh38
rs752173700 Chr.20: 37386199-37386199 on Build GRCh38
rs529267520 Chr.20: 37386062-37386062 on Build GRCh38
rs757241702 Chr.20: 37386100-37386100 on Build GRCh38
rs762030353 Chr.20: 37386174-37386174 on Build GRCh38
rs768964838 Chr.20: 37386160-37386160 on Build GRCh38
rs749015248 Chr.20: 37386097-37386097 on Build GRCh38
rs924953753 Chr.20: 37386040-37386040 on Build GRCh38
rs923008822 Chr.20: 37386106-37386106 on Build GRCh38
rs540214618 Chr.20: 37386022-37386022 on Build GRCh38
rs563936117 Chr.20: 37385989-37385989 on Build GRCh38
rs200926755 Chr.20: 37386049-37386049 on Build GRCh38
rs773537573 Chr.20: 37386182-37386182 on Build GRCh38

View Full Product Details