Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 306549-306810 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIER2
Entrez Gene ID
54531
Gene Name
MIER family member 2
Gene Aliases
KIAA1193, Mi-er2
UniGene
Hs.101891
Gene Chromosome Location
Chr.19: 305573-344793 on Build GRCh38
Transcripts
NM_017550.1 XM_005259582.4 XM_006722769.3 XM_011528072.2 XM_011528073.1 XM_011528074.2 XM_011528075.2 XM_011528077.2 XM_011528081.2 XM_011528082.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4607702 c.1635C>A p.C545* Chr.19: 306693-306693 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs200653769 Chr.19: 306671-306671 on Build GRCh38
rs1002948598 Chr.19: 306696-306696 on Build GRCh38
rs368468575 Chr.19: 306742-306742 on Build GRCh38
rs375042206 Chr.19: 306705-306705 on Build GRCh38
rs370296227 Chr.19: 306640-306640 on Build GRCh38
rs920191419 Chr.19: 306604-306604 on Build GRCh38
rs992192601 Chr.19: 306599-306599 on Build GRCh38
rs941354370 Chr.19: 306634-306634 on Build GRCh38
rs533062312 Chr.19: 306785-306785 on Build GRCh38
rs374520484 Chr.19: 306641-306641 on Build GRCh38
rs1040186035 Chr.19: 306764-306764 on Build GRCh38
rs753571661 Chr.19: 306678-306678 on Build GRCh38
rs1050737659 Chr.19: 306668-306668 on Build GRCh38
rs774524710 Chr.19: 306681-306681 on Build GRCh38
rs368456939 Chr.19: 306723-306723 on Build GRCh38
rs757823596 Chr.19: 306687-306687 on Build GRCh38
rs371479412 Chr.19: 306762-306762 on Build GRCh38
rs377758844 Chr.19: 306571-306571 on Build GRCh38
rs750719634 Chr.19: 306715-306715 on Build GRCh38
rs907601610 Chr.19: 306650-306650 on Build GRCh38
rs117553353 Chr.19: 306740-306740 on Build GRCh38
rs748295305 Chr.19: 306743-306743 on Build GRCh38
rs759081153 Chr.19: 306679-306679 on Build GRCh38
rs779453250 Chr.19: 306571-306571 on Build GRCh38
rs983236299 Chr.19: 306620-306620 on Build GRCh38
rs761742967 Chr.19: 306666-306666 on Build GRCh38
rs1040048806 Chr.19: 306654-306654 on Build GRCh38
rs774231082 Chr.19: 306665-306665 on Build GRCh38
rs201803724 Chr.19: 306652-306652 on Build GRCh38
rs910000090 Chr.19: 306632-306632 on Build GRCh38
rs917396256 Chr.19: 306597-306597 on Build GRCh38
rs928671241 Chr.19: 306575-306575 on Build GRCh38
rs372021700 Chr.19: 306737-306737 on Build GRCh38
rs201491939 Chr.19: 306675-306675 on Build GRCh38
rs576544501 Chr.19: 306739-306740 on Build GRCh38
rs372284005 Chr.19: 306667-306667 on Build GRCh38
rs369826991 Chr.19: 306786-306786 on Build GRCh38
rs551671742 Chr.19: 306787-306787 on Build GRCh38
rs958940808 Chr.19: 306598-306598 on Build GRCh38
rs772881235 Chr.19: 306672-306672 on Build GRCh38
rs534618782 Chr.19: 306680-306680 on Build GRCh38
rs780090477 Chr.19: 306728-306728 on Build GRCh38
rs760560149 Chr.19: 306674-306674 on Build GRCh38
rs557571212 Chr.19: 306702-306702 on Build GRCh38
rs958915898 Chr.19: 306722-306722 on Build GRCh38
rs368519912 Chr.19: 306647-306647 on Build GRCh38
rs951526960 Chr.19: 306612-306612 on Build GRCh38
rs367661892 Chr.19: 306754-306754 on Build GRCh38
rs992986032 Chr.19: 306585-306585 on Build GRCh38
rs369889630 Chr.19: 306778-306778 on Build GRCh38
rs749542521 Chr.19: 306736-306736 on Build GRCh38
rs763614445 Chr.19: 306780-306780 on Build GRCh38
rs566873957 Chr.19: 306570-306570 on Build GRCh38
rs138670550 Chr.19: 306753-306753 on Build GRCh38
rs865851183 Chr.19: 306635-306635 on Build GRCh38
rs569486833 Chr.19: 306788-306788 on Build GRCh38

View Full Product Details