Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 35259077-35259350 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLFN5
Entrez Gene ID
162394
Gene Name
schlafen family member 5
Gene Aliases
-
UniGene
Hs.709347
Gene Chromosome Location
Chr.17: 35243026-35273650 on Build GRCh38
Transcripts
NM_144975.3 XM_005257934.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6146140 c.533G>T p.R178I Chr.17: 35259223-35259223 on Build GRCh38
7140127 c.600C>A p.F200L Chr.17: 35259290-35259290 on Build GRCh38
5464003 c.492T>C p.Y164Y Chr.17: 35259182-35259182 on Build GRCh38
5464002 c.415T>G p.C139G Chr.17: 35259105-35259105 on Build GRCh38
6146139 c.603G>A p.S201S Chr.17: 35259293-35259293 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780386441 Chr.17: 35259187-35259187 on Build GRCh38
rs143657402 Chr.17: 35259125-35259125 on Build GRCh38
rs149039951 Chr.17: 35259310-35259310 on Build GRCh38
rs201606227 Chr.17: 35259109-35259109 on Build GRCh38
rs1041583366 Chr.17: 35259162-35259162 on Build GRCh38
rs781516346 Chr.17: 35259244-35259244 on Build GRCh38
rs756308518 Chr.17: 35259178-35259178 on Build GRCh38
rs115407444 Chr.17: 35259299-35259299 on Build GRCh38
rs751884807 Chr.17: 35259247-35259247 on Build GRCh38
rs770925097 Chr.17: 35259217-35259217 on Build GRCh38
rs746973579 Chr.17: 35259215-35259215 on Build GRCh38
rs564913392 Chr.17: 35259286-35259286 on Build GRCh38
rs752112206 Chr.17: 35259134-35259134 on Build GRCh38
rs774714975 Chr.17: 35259312-35259312 on Build GRCh38
rs143423216 Chr.17: 35259160-35259160 on Build GRCh38
rs776018361 Chr.17: 35259224-35259224 on Build GRCh38
rs999754149 Chr.17: 35259324-35259324 on Build GRCh38
rs773090779 Chr.17: 35259211-35259211 on Build GRCh38
rs202068652 Chr.17: 35259311-35259311 on Build GRCh38
rs760664870 Chr.17: 35259098-35259098 on Build GRCh38
rs766008815 Chr.17: 35259108-35259108 on Build GRCh38
rs762574342 Chr.17: 35259238-35259238 on Build GRCh38
rs935804945 Chr.17: 35259191-35259191 on Build GRCh38
rs376484895 Chr.17: 35259251-35259251 on Build GRCh38
rs139273106 Chr.17: 35259228-35259228 on Build GRCh38
rs762228016 Chr.17: 35259319-35259319 on Build GRCh38
rs376906702 Chr.17: 35259295-35259295 on Build GRCh38
rs764886677 Chr.17: 35259123-35259123 on Build GRCh38
rs993178457 Chr.17: 35259287-35259287 on Build GRCh38
rs755208700 Chr.17: 35259292-35259292 on Build GRCh38
rs150596834 Chr.17: 35259124-35259124 on Build GRCh38
rs767739047 Chr.17: 35259328-35259328 on Build GRCh38
rs867979867 Chr.17: 35259101-35259101 on Build GRCh38
rs144937987 Chr.17: 35259302-35259302 on Build GRCh38
rs768015031 Chr.17: 35259262-35259262 on Build GRCh38
rs753837575 Chr.17: 35259198-35259198 on Build GRCh38
rs764649766 Chr.17: 35259229-35259229 on Build GRCh38
rs1054405645 Chr.17: 35259220-35259220 on Build GRCh38
rs1018312221 Chr.17: 35259307-35259307 on Build GRCh38
rs764413036 Chr.17: 35259133-35259133 on Build GRCh38
rs764466457 Chr.17: 35259241-35259241 on Build GRCh38
rs745897225 Chr.17: 35259177-35259177 on Build GRCh38
rs754132339 Chr.17: 35259290-35259290 on Build GRCh38
rs756254815 Chr.17: 35259282-35259282 on Build GRCh38
rs750885614 Chr.17: 35259272-35259272 on Build GRCh38
rs752152324 Chr.17: 35259114-35259114 on Build GRCh38
rs199778009 Chr.17: 35259168-35259168 on Build GRCh38
rs775249106 Chr.17: 35259235-35259235 on Build GRCh38
rs777714710 Chr.17: 35259293-35259293 on Build GRCh38
rs746742608 Chr.17: 35259294-35259294 on Build GRCh38
rs769396858 Chr.17: 35259300-35259300 on Build GRCh38
rs148155981 Chr.17: 35259188-35259188 on Build GRCh38
rs759115370 Chr.17: 35259119-35259119 on Build GRCh38

View Full Product Details