Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 36033551-36033802 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPRD1A
Entrez Gene ID
55197
Gene Name
regulation of nuclear pre-mRNA domain containing 1A
Gene Aliases
HsT3101, P15RS
UniGene
Hs.464912
Gene Chromosome Location
Chr.18: 35982385-36067594 on Build GRCh38
Transcripts
NM_001303411.1 NM_001303412.1 NM_001303413.1 NM_018170.4 XM_011526048.1 XM_011526049.1 XM_011526050.1 XM_017025808.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
673857 c.262G>A p.A88T Chr.18: 36033727-36033727 on Build GRCh38
5028432 c.233C>G p.T78R Chr.18: 36033756-36033756 on Build GRCh38
4870637 c.101G>T p.S34I Chr.18: 36033780-36033780 on Build GRCh38
988187 c.209G>T p.S70I Chr.18: 36033780-36033780 on Build GRCh38
4072030 c.264T>C p.A88A Chr.18: 36033725-36033725 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746008904 Chr.18: 36033697-36033698 on Build GRCh38
rs554793433 Chr.18: 36033613-36033613 on Build GRCh38
rs781389048 Chr.18: 36033683-36033684 on Build GRCh38
rs747533361 Chr.18: 36033700-36033700 on Build GRCh38
rs770442162 Chr.18: 36033737-36033737 on Build GRCh38
rs749506360 Chr.18: 36033661-36033661 on Build GRCh38
rs755181268 Chr.18: 36033664-36033664 on Build GRCh38
rs897472561 Chr.18: 36033736-36033736 on Build GRCh38
rs3737474 Chr.18: 36033632-36033632 on Build GRCh38
rs564757717 Chr.18: 36033618-36033622 on Build GRCh38
rs111706297 Chr.18: 36033754-36033754 on Build GRCh38
rs1028905977 Chr.18: 36033742-36033742 on Build GRCh38
rs770249101 Chr.18: 36033758-36033758 on Build GRCh38
rs901685813 Chr.18: 36033655-36033655 on Build GRCh38
rs771058004 Chr.18: 36033682-36033682 on Build GRCh38
rs779384123 Chr.18: 36033667-36033667 on Build GRCh38
rs776052941 Chr.18: 36033740-36033740 on Build GRCh38
rs538014611 Chr.18: 36033610-36033610 on Build GRCh38
rs201716759 Chr.18: 36033697-36033697 on Build GRCh38
rs748533057 Chr.18: 36033676-36033676 on Build GRCh38
rs748628112 Chr.18: 36033687-36033687 on Build GRCh38
rs780038599 Chr.18: 36033657-36033657 on Build GRCh38
rs772645009 Chr.18: 36033689-36033689 on Build GRCh38
rs1044844940 Chr.18: 36033599-36033599 on Build GRCh38
rs746928555 Chr.18: 36033674-36033675 on Build GRCh38
rs1003641449 Chr.18: 36033686-36033686 on Build GRCh38

View Full Product Details