Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 102872946-102873179 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MMP12
Entrez Gene ID
4321
Gene Name
matrix metallopeptidase 12
Gene Aliases
HME, ME, MME, MMP-12
UniGene
Hs.1695
Gene Chromosome Location
Chr.11: 102862729-102875034 on Build GRCh38
Transcripts
NM_002426.5

SNP Detail

SNP ID Chromosome Location
rs200528927 Chr.11: 102873123-102873123 on Build GRCh38
rs972827323 Chr.11: 102873045-102873045 on Build GRCh38
rs200208956 Chr.11: 102873102-102873102 on Build GRCh38
rs201783229 Chr.11: 102873147-102873147 on Build GRCh38
rs781874658 Chr.11: 102873059-102873059 on Build GRCh38
rs782550942 Chr.11: 102873013-102873013 on Build GRCh38
rs200326291 Chr.11: 102873051-102873051 on Build GRCh38
rs782223469 Chr.11: 102873114-102873114 on Build GRCh38
rs201533418 Chr.11: 102873129-102873129 on Build GRCh38
rs373941993 Chr.11: 102872986-102872986 on Build GRCh38
rs782039320 Chr.11: 102872974-102872974 on Build GRCh38
rs372705906 Chr.11: 102873128-102873128 on Build GRCh38
rs201876594 Chr.11: 102873085-102873085 on Build GRCh38
rs200900481 Chr.11: 102873081-102873081 on Build GRCh38
rs368045839 Chr.11: 102872990-102872990 on Build GRCh38
rs552593852 Chr.11: 102873116-102873117 on Build GRCh38
rs201266907 Chr.11: 102873036-102873036 on Build GRCh38
rs782011685 Chr.11: 102873109-102873109 on Build GRCh38
rs201052180 Chr.11: 102873106-102873106 on Build GRCh38
rs962766794 Chr.11: 102872982-102872982 on Build GRCh38
rs200085197 Chr.11: 102872992-102872992 on Build GRCh38
rs199627800 Chr.11: 102873117-102873117 on Build GRCh38
rs782049815 Chr.11: 102873117-102873117 on Build GRCh38
rs782141106 Chr.11: 102873127-102873127 on Build GRCh38
rs534882108 Chr.11: 102873138-102873138 on Build GRCh38
rs950104425 Chr.11: 102873091-102873091 on Build GRCh38
rs782437708 Chr.11: 102873149-102873149 on Build GRCh38
rs200813655 Chr.11: 102873152-102873152 on Build GRCh38
rs782467921 Chr.11: 102873065-102873065 on Build GRCh38
rs782334342 Chr.11: 102873024-102873025 on Build GRCh38
rs371800506 Chr.11: 102873067-102873067 on Build GRCh38
rs782451057 Chr.11: 102873090-102873090 on Build GRCh38
rs781964741 Chr.11: 102872973-102872973 on Build GRCh38
rs781889708 Chr.11: 102873154-102873154 on Build GRCh38
rs782677013 Chr.11: 102873098-102873098 on Build GRCh38
rs200664069 Chr.11: 102873141-102873141 on Build GRCh38
rs202180058 Chr.11: 102873038-102873038 on Build GRCh38
rs782568490 Chr.11: 102873155-102873155 on Build GRCh38
rs140752089 Chr.11: 102873137-102873137 on Build GRCh38
rs376435056 Chr.11: 102873104-102873104 on Build GRCh38
rs781879770 Chr.11: 102873000-102873000 on Build GRCh38
rs781928619 Chr.11: 102873122-102873122 on Build GRCh38
rs150120347 Chr.11: 102873150-102873150 on Build GRCh38

View Full Product Details