Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 2942119-2942392 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOP14
Entrez Gene ID
8602
Gene Name
NOP14 nucleolar protein
Gene Aliases
C4orf9, NOL14, RES4-25, RES425, UTP2
UniGene
Hs.627133
Gene Chromosome Location
Chr.4: 2937936-2963506 on Build GRCh38
Transcripts
NM_001291978.1 NM_001291979.1 NM_003703.2 NR_015453.2

Gene Symbol
NOP14-AS1
Entrez Gene ID
317648
Gene Name
NOP14 antisense RNA 1
Gene Aliases
C4orf10, RES4-24
UniGene
Hs.398178
Gene Chromosome Location
Chr.4: 2935546-2951078 on Build GRCh38
Transcripts
NM_001291978.1 NM_001291979.1 NM_003703.2 NR_015453.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1310010 c.1996C>G p.L666V Chr.4: 2942247-2942247 on Build GRCh38
6727778 c.2031G>A p.S677S Chr.4: 2942212-2942212 on Build GRCh38
733622 c.1935G>A p.S645S Chr.4: 2942308-2942308 on Build GRCh38
3603289 c.1998C>T p.L666L Chr.4: 2942245-2942245 on Build GRCh38
6836280 c.1954G>C p.A652P Chr.4: 2942289-2942289 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758688143 Chr.4: 2942183-2942183 on Build GRCh38
rs773125467 Chr.4: 2942150-2942150 on Build GRCh38
rs145795270 Chr.4: 2942317-2942317 on Build GRCh38
rs376483192 Chr.4: 2942156-2942156 on Build GRCh38
rs770714351 Chr.4: 2942228-2942228 on Build GRCh38
rs149885863 Chr.4: 2942270-2942270 on Build GRCh38
rs376705776 Chr.4: 2942348-2942348 on Build GRCh38
rs370106585 Chr.4: 2942349-2942349 on Build GRCh38
rs202046753 Chr.4: 2942269-2942269 on Build GRCh38
rs370425099 Chr.4: 2942151-2942151 on Build GRCh38
rs771639187 Chr.4: 2942253-2942253 on Build GRCh38
rs752789508 Chr.4: 2942294-2942294 on Build GRCh38
rs767879615 Chr.4: 2942357-2942357 on Build GRCh38
rs139007665 Chr.4: 2942259-2942259 on Build GRCh38
rs759874022 Chr.4: 2942350-2942350 on Build GRCh38
rs768154236 Chr.4: 2942340-2942340 on Build GRCh38
rs995458523 Chr.4: 2942177-2942177 on Build GRCh38
rs376246926 Chr.4: 2942192-2942192 on Build GRCh38
rs373247234 Chr.4: 2942244-2942244 on Build GRCh38
rs184398419 Chr.4: 2942366-2942366 on Build GRCh38
rs770454368 Chr.4: 2942152-2942152 on Build GRCh38
rs758994754 Chr.4: 2942160-2942160 on Build GRCh38
rs759333863 Chr.4: 2942276-2942277 on Build GRCh38
rs764170907 Chr.4: 2942367-2942367 on Build GRCh38
rs372098273 Chr.4: 2942324-2942324 on Build GRCh38
rs1036606875 Chr.4: 2942168-2942168 on Build GRCh38
rs370813940 Chr.4: 2942310-2942310 on Build GRCh38
rs767752259 Chr.4: 2942289-2942289 on Build GRCh38
rs373019587 Chr.4: 2942175-2942175 on Build GRCh38
rs767226195 Chr.4: 2942278-2942279 on Build GRCh38
rs778767912 Chr.4: 2942308-2942308 on Build GRCh38
rs925740421 Chr.4: 2942280-2942280 on Build GRCh38
rs766698228 Chr.4: 2942284-2942284 on Build GRCh38
rs77265212 Chr.4: 2942323-2942323 on Build GRCh38
rs773891316 Chr.4: 2942243-2942243 on Build GRCh38
rs1019020511 Chr.4: 2942257-2942257 on Build GRCh38
rs111772006 Chr.4: 2942328-2942328 on Build GRCh38
rs376428719 Chr.4: 2942276-2942276 on Build GRCh38
rs200567683 Chr.4: 2942369-2942369 on Build GRCh38
rs149018189 Chr.4: 2942298-2942298 on Build GRCh38
rs200033536 Chr.4: 2942208-2942208 on Build GRCh38
rs991518719 Chr.4: 2942143-2942143 on Build GRCh38
rs373966810 Chr.4: 2942247-2942247 on Build GRCh38
rs151173699 Chr.4: 2942238-2942238 on Build GRCh38
rs752663755 Chr.4: 2942281-2942282 on Build GRCh38
rs558519566 Chr.4: 2942182-2942182 on Build GRCh38
rs763594492 Chr.4: 2942207-2942207 on Build GRCh38
rs766958962 Chr.4: 2942162-2942162 on Build GRCh38
rs777597555 Chr.4: 2942296-2942296 on Build GRCh38
rs781415421 Chr.4: 2942197-2942197 on Build GRCh38
rs749072590 Chr.4: 2942212-2942212 on Build GRCh38
rs753787388 Chr.4: 2942371-2942371 on Build GRCh38
rs908292793 Chr.4: 2942302-2942302 on Build GRCh38
rs760431479 Chr.4: 2942299-2942299 on Build GRCh38
rs751793865 Chr.4: 2942285-2942285 on Build GRCh38
rs773596681 Chr.4: 2942341-2942341 on Build GRCh38
rs763510588 Chr.4: 2942345-2942345 on Build GRCh38
rs748151163 Chr.4: 2942205-2942205 on Build GRCh38
rs145858265 Chr.4: 2942287-2942287 on Build GRCh38
rs144256817 Chr.4: 2942193-2942193 on Build GRCh38
rs774970141 Chr.4: 2942258-2942258 on Build GRCh38
rs756217921 Chr.4: 2942295-2942295 on Build GRCh38
rs995111066 Chr.4: 2942330-2942330 on Build GRCh38
rs777129498 Chr.4: 2942174-2942174 on Build GRCh38
rs548054366 Chr.4: 2942236-2942236 on Build GRCh38
rs775000247 Chr.4: 2942255-2942255 on Build GRCh38
rs746553674 Chr.4: 2942326-2942326 on Build GRCh38

View Full Product Details