Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49968298-49968567 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBM6
Entrez Gene ID
10180
Gene Name
RNA binding motif protein 6
Gene Aliases
3G2, DEF-3, DEF3, HLC-11, NY-LU-12, g16
UniGene
Hs.696735
Gene Chromosome Location
Chr.3: 49940038-50077252 on Build GRCh38
Transcripts
NM_001167582.1 NM_005777.2 XM_005264784.1 XM_005264785.1 XM_005264786.1 XM_005264787.2 XM_006712916.1 XM_017005496.1 XM_017005498.1 XM_017005499.1 XM_017005500.1 XM_017005501.1 XM_017005502.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3427715 c.1022A>G p.E341G Chr.3: 49968447-49968447 on Build GRCh38
5014344 c.1060C>G p.P354A Chr.3: 49968485-49968485 on Build GRCh38
6039468 c.1020_1022delAGA p.E341delE Chr.3: 49968445-49968447 on Build GRCh38
4118682 c.1118C>T p.S373F Chr.3: 49968543-49968543 on Build GRCh38
7110850 c.930G>A p.V310V Chr.3: 49968355-49968355 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763240756 Chr.3: 49968390-49968390 on Build GRCh38
rs1028110367 Chr.3: 49968387-49968387 on Build GRCh38
rs756028153 Chr.3: 49968433-49968433 on Build GRCh38
rs755207084 Chr.3: 49968412-49968412 on Build GRCh38
rs751789723 Chr.3: 49968399-49968399 on Build GRCh38
rs771713587 Chr.3: 49968505-49968505 on Build GRCh38
rs373537065 Chr.3: 49968430-49968430 on Build GRCh38
rs61731329 Chr.3: 49968483-49968483 on Build GRCh38
rs767321378 Chr.3: 49968421-49968421 on Build GRCh38
rs370873930 Chr.3: 49968459-49968459 on Build GRCh38
rs770906778 Chr.3: 49968463-49968463 on Build GRCh38
rs762254968 Chr.3: 49968344-49968344 on Build GRCh38
rs1035936547 Chr.3: 49968408-49968408 on Build GRCh38
rs745544778 Chr.3: 49968492-49968492 on Build GRCh38
rs765679845 Chr.3: 49968378-49968378 on Build GRCh38
rs374241916 Chr.3: 49968515-49968515 on Build GRCh38
rs930854245 Chr.3: 49968545-49968545 on Build GRCh38
rs770294018 Chr.3: 49968518-49968518 on Build GRCh38
rs759384829 Chr.3: 49968414-49968414 on Build GRCh38
rs953762761 Chr.3: 49968398-49968398 on Build GRCh38
rs777786423 Chr.3: 49968436-49968436 on Build GRCh38
rs775030359 Chr.3: 49968337-49968337 on Build GRCh38
rs766728138 Chr.3: 49968396-49968396 on Build GRCh38
rs977439394 Chr.3: 49968447-49968447 on Build GRCh38
rs774881304 Chr.3: 49968511-49968511 on Build GRCh38
rs771529136 Chr.3: 49968332-49968332 on Build GRCh38
rs750804570 Chr.3: 49968388-49968388 on Build GRCh38
rs759211587 Chr.3: 49968329-49968329 on Build GRCh38
rs763440033 Chr.3: 49968532-49968532 on Build GRCh38
rs773779624 Chr.3: 49968529-49968529 on Build GRCh38
rs143972186 Chr.3: 49968422-49968422 on Build GRCh38
rs200888268 Chr.3: 49968354-49968354 on Build GRCh38
rs972734908 Chr.3: 49968438-49968438 on Build GRCh38
rs1003574576 Chr.3: 49968391-49968391 on Build GRCh38

View Full Product Details