Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 88459690-88459915 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MRPL46
Entrez Gene ID
26589
Gene Name
mitochondrial ribosomal protein L46
Gene Aliases
C15orf4, LIECG2, P2ECSL
UniGene
Hs.534261
Gene Chromosome Location
Chr.15: 88459478-88467402 on Build GRCh38
Transcripts
NM_022163.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5121038 c.605C>G p.A202G Chr.15: 88459848-88459848 on Build GRCh38
3361569 c.729T>C p.F243F Chr.15: 88459724-88459724 on Build GRCh38
5187192 c.627C>T p.P209P Chr.15: 88459826-88459826 on Build GRCh38
2014476 c.755A>G p.H252R Chr.15: 88459698-88459698 on Build GRCh38
346439 c.665G>T p.R222L Chr.15: 88459788-88459788 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779860260 Chr.15: 88459857-88459857 on Build GRCh38
rs368264703 Chr.15: 88459845-88459845 on Build GRCh38
rs761057525 Chr.15: 88459876-88459877 on Build GRCh38
rs544918347 Chr.15: 88459824-88459824 on Build GRCh38
rs771535053 Chr.15: 88459887-88459887 on Build GRCh38
rs564581312 Chr.15: 88459803-88459803 on Build GRCh38
rs775939549 Chr.15: 88459838-88459841 on Build GRCh38
rs774704451 Chr.15: 88459751-88459753 on Build GRCh38
rs1000988299 Chr.15: 88459727-88459727 on Build GRCh38
rs766716247 Chr.15: 88459739-88459739 on Build GRCh38
rs367735118 Chr.15: 88459798-88459798 on Build GRCh38
rs772592906 Chr.15: 88459808-88459808 on Build GRCh38
rs752852980 Chr.15: 88459771-88459771 on Build GRCh38
rs745858607 Chr.15: 88459781-88459781 on Build GRCh38
rs1021164035 Chr.15: 88459712-88459712 on Build GRCh38
rs781290999 Chr.15: 88459774-88459774 on Build GRCh38
rs769680679 Chr.15: 88459785-88459785 on Build GRCh38
rs140277766 Chr.15: 88459789-88459789 on Build GRCh38
rs768299497 Chr.15: 88459868-88459868 on Build GRCh38
rs778527807 Chr.15: 88459871-88459871 on Build GRCh38
rs113945322 Chr.15: 88459726-88459726 on Build GRCh38
rs778999138 Chr.15: 88459765-88459765 on Build GRCh38
rs766741187 Chr.15: 88459806-88459806 on Build GRCh38
rs750731317 Chr.15: 88459721-88459721 on Build GRCh38
rs1037863891 Chr.15: 88459859-88459859 on Build GRCh38
rs149923755 Chr.15: 88459860-88459860 on Build GRCh38
rs765381395 Chr.15: 88459819-88459819 on Build GRCh38
rs779764453 Chr.15: 88459787-88459787 on Build GRCh38
rs754055880 Chr.15: 88459741-88459741 on Build GRCh38
rs373706267 Chr.15: 88459792-88459792 on Build GRCh38
rs142867191 Chr.15: 88459713-88459713 on Build GRCh38
rs746177582 Chr.15: 88459762-88459762 on Build GRCh38
rs151073015 Chr.15: 88459772-88459772 on Build GRCh38
rs771632952 Chr.15: 88459797-88459797 on Build GRCh38
rs761017773 Chr.15: 88459735-88459735 on Build GRCh38
rs756029941 Chr.15: 88459855-88459855 on Build GRCh38
rs531998570 Chr.15: 88459818-88459818 on Build GRCh38
rs747867213 Chr.15: 88459880-88459880 on Build GRCh38
rs768020076 Chr.15: 88459716-88459716 on Build GRCh38
rs758539499 Chr.15: 88459833-88459833 on Build GRCh38
rs755201593 Chr.15: 88459763-88459763 on Build GRCh38
rs983318117 Chr.15: 88459796-88459796 on Build GRCh38
rs572609825 Chr.15: 88459788-88459788 on Build GRCh38
rs754174601 Chr.15: 88459811-88459811 on Build GRCh38
rs749043815 Chr.15: 88459865-88459865 on Build GRCh38

View Full Product Details